Skip to Content
Merck
All Photos(3)

Documents

EP6333000014

Eppendorf® Mastercycler® Nexus Thermal Cyclers

new

complete unit, aluminum block, AC/DC input 230 V, 50—60 Hz, EuroPlug

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41102422
UNSPSC Code:
41102421

packaging

pack of 1 ea

manufacturer/tradename

Eppendorf® 6333000014

Looking for similar products? Visit Product Comparison Guide

General description

Mastercycler nexus 230 V/ 50-60 Hz
Product Information
With the versatile Mastercycler nexus you can choose the PCR consumables that best suit your assay. The PCR cycler nexus can accommodate 96-well PCR plates, 0.2 mL PCR tubes, 0.2 mL PCR tube strips and 0.5 mL PCR tubes. With a noise level of less than 40 dB[a], the nexus is very quiet and energy-efficient at the same time.

In the age of networking, the new Mastercycler nexus is your reliable companion when it comes to the daily routine of PCR. It is easy to use, does not need much space or energy and sends you an e-mail when it is done – what else do you need from a PCR cycler?

Certified Quality
All Eppendorf cyclers follow these quality guidelines:


  • Individual, documented quality control certificates
  • Calibration accordingly to national and international standards: NIST (USA), DKD/PTB (Germany), UKAS/NPL (UK)
  • UL/cUL listed

The Mastercycler nexus is a whisper quiet PCR machine. Low noise levels make working in your lab more pleasant and enjoyable, particularly when you are thinking about your next big breakthrough findings….

Application

Standard PCR, Fast PCR, PCR optimization

Features and Benefits

  • Intuitive graphic programming
  • Small footprint
  • Universal block for ultimate flexibility
  • Connect up to 2 Mastercycler nexus eco to your existing unit
  • Available with gradient
  • E-mail notification
  • Flexlid® concept allows use of all types of consumables with automatic height adjustment of the lid

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Mastercycler is a registered trademark of Eppendorf AG
flexlid is a registered trademark of Eppendorf AG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service