Key Documents
EP6001081110
Eppendorf® Side Access Stainless Steel Rack for CryoCube(R) F740 Upright Freezers
including front and back handle for storage of 36 Deepwell plates up to 86x128x53 mm, 5-Compartment, external W × D × H (139 mm) (569 mm) (343 mm)
About This Item
UNSPSC Code:
41122800
material
stainless steel
feature
5-Compartment
Holds 48 x deepwell plates
including front and back handle for storage of 36 Deepwell plates up to 86x128x53 mm
manufacturer/tradename
Eppendorf® 6001081110
technique(s)
cryopreservation: suitable
external W × D × H
(139 mm) (569 mm) (343 mm)
size
549x139x343 mm
General description
Side access style stainless steel racks for Eppendorf CryoCube F740 ultra-low temperature upright freezers. By storing your sample inventory in an organized manner, you enable more economical use of the available space in your ULT freezer. Racks allow you to use 100% of the storage capacity. When using racks with side access, most of the storage boxes have to be pulled out of the cold interior of the ultra-low temperature freezer when accessing samples. In addition, metal racks have a long lifespan and can often be used for the next generation of ULT freezers. The stainless steel racks are assembled with 100% energy from renewable sources and made from 60-65% recycled steel. Coupled with Eppendorf′s use of cardboard packaging made from 90% recycled raw materials these are a good option to reduce your carbon footprint.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)