Key Documents
EP5895118003
Rotor for Eppendorf® 5920R Centrifuges
swing-bucket rotor, S-4x1000, non aerosol tight, to hold, 8 x MTP/DWP plates/tubes, with 4 high-capacity 1000 mL buckets
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
Product EP5895118003 is not currently sold in your country. Contact Technical Service
feature
to hold 8 x MTP/DWP plates/tubes
non aerosol tight
swing-bucket rotor
with 4 high-capacity 1000 mL buckets
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5895118003 (Global)
parameter
3,153 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5920R
General description
The large Centrifuge 5920 R is a benchtop centrifuge that combines exceptional capacity with a compact and ergonomic design. It has similar dimensions to refrigerated competitor models in the 3-liter segment, but has an outstanding capacity of up to 4 x 1,000 mL or 52 x 50 mL tubes. Optimize high-throughput centrifugation and make your daily work easier.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)