Skip to Content
Merck
All Photos(1)

Key Documents

EP5820711000

Eppendorf® Bucket for Deep-Well Plates

used with Rotor A-2-DWP-AT

Synonym(s):

Eppendorf® Bucket for Deep-Well Plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103910
NACRES:
NB.15
Product EP5820711000 is not currently sold in your country. Contact Technical Service

packaging

pack of 2 ea

manufacturer/tradename

Eppendorf® 5820711000

technique(s)

centrifugation: suitable

compatibility

used with Rotor A-2-DWP-AT

General description

Spare buckets Rotor A-2-DWP-AT set of 2

Application

CENTRIFUGATION

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service