Skip to Content
Merck
All Photos(1)

Key Documents

EP5703000320

Eppendorf® Centrifuge 5702 R G

incl. rotor A-4-38 and 13/16 mL adapters, AC/DC input 230 V, 50 - 60 Hz

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161701
NACRES:
NB.81
Product EP5703000320 is not currently sold in your country. Contact Technical Service

AC/DC input

230 V AC, Schuko plug

feature

CE compliant
refrigerated
to hold 16 13/16 mm blood collection tubes
to hold 4 100 mL round buckets
incl. rotor A-4-38 and 13/16 mL adapters

packaging

pkg of 1 ea

manufacturer/tradename

Eppendorf® 5703000320 (EU)

parameter

temp. range at -9-40 °C
3000 × g
4400 rpm

technique(s)

centrifugation: suitable

Looking for similar products? Visit Product Comparison Guide

General description

The 5702 family of low speed, general purpose centrifuges was especially designed to meet the requirement of low- to mid-throughput research and cell culture labs. They are compact in design so that one can fit comfortably on every lab bench, quiet in operation to benefit your work environment, and provide 6 different rotor options to accommodate any type of tube.
  • Centrifuge 5702 for standard applications
  • Refrigerated Centrifuge 5702 R for heat sensitive samples


Quality and safety

  • Stainless steel rotor chamber is rust-free and easy to clean
  • Imbalance detection for maximum safety
  • Key lock to prevent accidental parameter changes
  • Aerosol-tight caps for safe centrifugation of hazardous samples optionally available

Features and Benefits

  • Max. speed: 3,000 × g (4,400 rpm)
  • Very compact footprint to fit on every lab bench
  • Very quiet operation to benefit your environment
  • Low access height for simple loading and unloading of samples
  • SOFT brake function for slow acceleration and braking. Optimized for cell separation by gradient centrifugation
  • “At set RPM” function starts the timer when the selected rotational speed is reached for reproducible centrifugation runs
  • Key lock to prevent unintentional adjustments
  • Stainless steel rotor chamber is rust-free and easy to clean
  • Electronic imbalance detection for maximum safety

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service