Key Documents
EP5452727007
Eppendorf® Rotor F-55-16-5-PCR
incl. rotor lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 2 x PCR strips
incl. rotor lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5452727007
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with centrifuge models MiniSpin/MiniSpin Plus
Legal Information
Eppendorf is a registered trademark of Eppendorf AG