Skip to Content
Merck
All Photos(1)

Key Documents

EP0030501241

Eppendorf® Deepwell plates, standard, 96 wells

blue plate, conical bottom colorless wells, capacity 1000 μL, pkg of 20 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.15
material:
blue plate
conical bottom colorless wells
size:
96 wells
sterility:
sterile; autoclaved
Product EP0030501241 is not currently sold in your country. Contact Technical Service

material

blue plate
conical bottom colorless wells

sterility

sterile; autoclaved

packaging

pkg of 20 ea

manufacturer/tradename

Eppendorf® 0030501241

availability

available only in (India)

technique(s)

PCR: suitable

capacity

1000 μL

size

96 wells

working volume

1 mL

Looking for similar products? Visit Product Comparison Guide

General description

For greater reliability, stability and efficiency, choose Eppendorf Deepwell 96 well Plates. Available with white, yellow, green or blue frames, these deepwell plates offer high performance for all applications - both manual and automated. Thanks to OptiTrack matrix, a high-contrast alphanumeric label laser-engraved into the frame, you can find your sample up to 30% faster and reduce pipetting errors. For every application, from sample storage at 86 C to DNA denaturation at 100 C, rely on Eppendorf Deepwell Plates and set new standards in your laboratory.

Application

  • Sample storage and preparation Bacteria and yeast cultivation
  • High-throughput DNA and RNA isolation
  • Storage of cDNA or genomic libraries
  • Storage of siRNA or oligonucleotide libraries
  • Protein analysis Storage of valuable protein and DNA samples in Eppendorf LoBind Plates
  • High-throughput plasmid purification Creation of dilution series

Features and Benefits

  • OptiTrack matrix: up to 30 % faster sample detection and fewer pipetting errors thanks to high-contrast alphanumeric labeling
  • RecoverMax well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
  • g-Safe : exceptional centrifugation resistance up to 6,000 × g for faster protocols and improved sample quality
  • Raised corrugated edges and smooth surface for reliable sealing, even during heat sealing
  • Easily and reliably stackable
  • Minimal residual volume and high homogeneity from well to well
  • High-quality polypropylene for high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without mold release agents, plasticizers and biocides – substances that can negatively affect bioassays
  • Autoclavable (121 C, 20 min)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service