EP0030129679
Eppendorf twin.tec® PCR Plates 96
PCR clean, skirted, yellow, pkg of 25 ea
About This Item
skirt
Recommended Products
material
yellow wells
sterility
non-sterile
feature
PCR clean skirted
skirt
packaging
pkg of 25 ea
manufacturer/tradename
Eppendorf® 0030129679
technique(s)
PCR: suitable
size
96 well
working volume
150 μL
color
yellow
binding type
low binding surface
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Maximum recovery rates for improved assay results
- Available with barcode (upon request)
- Ideal for quantitative real-time PCR as well
- Exceptionally solid and torque-resistant polycarbonate frame
- OptiTrack® matrix for faster sample identification and fewer pipetting errors
- Raised well rims for effective sealing, also reduces risk of cross-contamination
- Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service