Skip to Content
Merck
All Photos(1)

Key Documents

EP0030129555

Eppendorf® twin.tec PCR Plate 96 LoBind

skirt, PCR clean, green, pkg of 300 pieces

Synonym(s):

Eppendorf® twin.tec® PCR Plate 96 LoBind

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.15
material:
green wells
size:
96 well
sterility:
non-sterile
binding type:
low binding surface
feature:
PCR clean
skirt
Product EP0030129555 is not currently sold in your country. Contact Technical Service

material

green wells

sterility

non-sterile

feature

PCR clean
skirt

packaging

pkg of 300 pieces

manufacturer/tradename

Eppendorf® 0030129555

technique(s)

PCR: suitable

size

96 well

color

green

binding type

low binding surface

Looking for similar products? Visit Product Comparison Guide

General description

Eppendorf® twin.tec® Eppendorf LoBind® PCR plates are specially designed to improve your polymerase chain reaction (PCR) results by increasing the yield of your target molecules. Thin-walled polypropylene wells with LoBind surfaces have low DNA binding properties, leading to reduced loss of target molecules and higher yields in PCR and other molecular assays.
Eppendorf′s twin.tec® Eppendorf LoBind® PCR plates are available in 96- and 384-well formats. A combination of special manufacturing technologies and selected polypropylene batches ensures that nearly 100 % of DNA and RNA molecules are recovered without the use of surface coatings, thereby eliminating the risk of sample contamination.

Application

Eppendorf® twin.tec® PCR Plate 96 LoBind is suitable for:
  • PCR with low template concentration
  • Real-time PCR with low sample concentration
  • Low-volume PCR/qPCR
  • NGS DNA library construction

Features and Benefits

  • Eppendorf LoBind® material assures maximum sample recovery for improved assay results
  • 96 well-skirted PCR plates contain an extremely solid and torque-resistant polycarbonate frame and polypropylene wells for optimum performance
  • Extremely thin-walled polypropylene wells provide superior thermal cycling performance
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised rim design: enable effective sealing to reduce the risk of cross-contamination
  • Eppendorf® twin.tec PCR Plates are free of DNA, DNAase, RNase, and PCR inhibitors (PCR clean)
  • Available with barcode (upon request)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
Eppendorf LoBind is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service