Key Documents
EP0030128940
Eppendorf® Twin tec PCR plates, 384 wells
yellow polypropylene plate, conical bottom colorless wells, pkg of 300 ea
About This Item
UNSPSC Code:
41106300
NACRES:
NB.15
material:
conical bottom colorless wells
yellow polypropylene plate
size:
384 wells
binding type:
low binding surface
feature:
skirt (stackable)
Product EP0030128940 is not currently sold in your country. Contact Technical Service
material
conical bottom colorless wells
yellow polypropylene plate
feature
skirt (stackable)
packaging
pkg of 300 ea
manufacturer/tradename
Eppendorf® 0030128940
technique(s)
PCR: suitable
maximum volume
-45 μL
size
384 wells
working volume
-40 μL
suitability
suitable for PCR
binding type
low binding surface
General description
The Eppendorf “twin.tec” PCR plates are designed for single-use PCR applications. They have a rigid polycarbonate frame that retains its shape even when exposed to high temperatures during the PCR cycle. The plate is designed to maintain dimensional stability, providing support for both the snug fit and sealing functions. Additionally, the raised rims of the plate further enhance effective sealing by forming a highly secure seal with the sealing material.
Application
“Eppendorf” Twin tec PCR plates are suitable for:
  • Standard PCR
  • Real-time PCR
  • High Throughput PCR
  • Small Volume Sample Handling
Features and Benefits
  • Eppendorf “twin.tec” PCR plate has a precise fit to the PCR cycler and a sturdy base enables tight sealing.
  • It is free of prevalent PCR contaminants such as DNA, DNAse, RNAse, and PCR inhibitors.
  • Skirt is designed to optimize automation and barcoding.
  • The eight holes in the skirt facilitate easy positioning and removal of the plate from the thermal cycler block.
  • Idea for use in 384-well thermocyclers.
  • Maximum volume per well is 40 µL.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)