Skip to Content
Merck
All Photos(1)

Documents

EP0030127722

Eppendorf® Serological Pipets, Sterile Grade

orange, capacity 10 mL, pack of 400 ea (4 bags × 100 pcs.)

Synonym(s):

Pipettes, Serological Pipets, pipettes, pipetting tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.04

material

orange

sterility

sterile

feature

DNA free
graduations
plugged

packaging

pack of 400 ea (4 bags × 100 pcs.)

manufacturer/tradename

Eppendorf® 0030127722 (Global)

capacity

10 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® Serological Pipets are manufactured from ultra-pure virgin polystyrene for optimal clarity.
  • Easy-to-read graduations for accurate volume determination.
  • Pipets are color-coded based on sizes for easy identification.
  • Sterility assurance level of 10-6.
  • Compatible with all mechanical and electronic pipette controllers - including Pipet Helper® or Easypet® 3.
  • Free of detectable pyrogens, DNA, RNase and DNase and certified non-cytotoxic.

Application

  • Transfer of liquids
  • Mixing suspensions
  • Combining reagents and chemical solutions
  • Transferring cells for empirical analysis or expansion
  • Layering reagents for creating higher density gradients

Legal Information

Easypet is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Pipet Helper is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service