Skip to Content
Merck
All Photos(1)

Key Documents

EP0030122160

Eppendorf® Conical Tubes

capacity 15 mL, sterile, pyrogen-, DNase-, RNase-, and DNA-free, pack of 500 ea (20 racks x 25 tubes)

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.22
Product EP0030122160 is not currently sold in your country. Contact Technical Service

material

colorless tube
conical bottom
high-density polyethylene cap
polypropylene tube
screw top cap
white cap

sterility

sterile

feature

graduations
neck thread External thread
pyrogen-, DNase-, RNase-, and DNA-free

packaging

pack of 500 ea (20 racks x 25 tubes)

manufacturer/tradename

Eppendorf® 0030122160

capacity

15 mL

tube diam.

17 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Eppendorf® conical tubes are manufactured from high-quality chemically resistant polypropylene (PP).
  • Ideal for most cell culture and cell biology applications.
  • High g-Safe centrifugation stability, and optimal sample and pellet visibility.
  • Ergonomic design - one-handed operation for easy opening/closing.
  • Free of detectable slip agents, plasticizers, and biocides ensure optimal sample integrity.
  • Pyrogen-, DNase-, RNase-, human and bacterial DNA-free for convenient and safe cell culture applications.
  • Cap with flattened and grooved sides minimizes contamination and evaporation risks.
  • A large writing area on the tube facilitates accurate labeling.
  • Compatible with centrifuge rotors, mixers, and shakers.

Application

  • Sample preparation in microbiology and molecular biology labs
  • Large volume storage of samples and solutions.
  • Preparation of mastermixes and buffers.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service