Skip to Content
Merck
All Photos(1)

Key Documents

EP0030121589

Eppendorf® Safe-Lock micro test tubes, Biopur®

capacity 1.5 mL, pack of 100 ea

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121702
NACRES:
NB.11
Product EP0030121589 is not currently sold in your country. Contact Technical Service

material

cap (plug seal)
colorless cap
colorless tube
conical bottom
polypropylene cap
polypropylene tube

description

PCR inhibitor free
sterile, individually packed

sterility

sterile

feature

ATP free
DNA free
RNase and DNase free
endotoxin free
graduations
neck thread Push fit
pyrogen free

packaging

pack of 100 ea

manufacturer/tradename

Eppendorf® 0030121589

capacity

1.5 mL

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Eppendorf® Safe-Lock tubes, Biopur® (colorless) protect the sample with excellent centrifugation stability, feature optimized sealing properties, and reliably prevent sample evaporation. The features include:
  • Hinged Eppendorf Safe-Lock lid prevents unintentional lid opening during incubation and storage.
  • Polypropylene provides high resistance to chemicals, mechanical stress and temperature extremes.
  • Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results, g-Safe®: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples Precise lid sealing for minimal evaporation rates during long-term storage
  • Large frosted lid and surface on the side for easy labelling
  • Guaranteed functionality from -86 °C to 100 °C
  • Autoclavable when open (121 °C, 20 min)
  • Available in Eppendorf Protein and DNA LoBind material for maximum recovery of valuable samples.


Using the Eppendorf 30,000 × g system comprising of Safe-Lock Tubes and Centrifuge 5430 R with high-speed rotor, the effect of relative centrifugal force (rcf), duration of centrifugation and the amount of isopropanol on the recovery rate of plasmid DNA from alcohol precipitation were investigated. All three factors play a role, with rcf having the greatest effect. By centrifuging the Eppendorf Safe-Lock Tubes at 30,000 × g, nearly 90% of DNA could be recovered from a 5 min centrifugation.

Legal Information

Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
g-Safe is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service