Skip to Content
Merck
All Photos(1)

Key Documents

EMU202201

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Phb

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTCATTTCCTCATCCCTTGGGTACAGAAACCAATTATCTTTGACTGCCGCTCTCGACCACGGAACGTACCAGTCATCACTGGTAGCAAAGACTTGCAGAACGTCAATATCACACTGCGAATCCTCTTCCGGCCGGTGGCCAGCCAGCTTCCTCGTATCTACACCAGCATTGGCGAGGACTATGATGAGCGGGTCCTGCCTTCTATCACCACAGAGATCCTCAAGTCGGTGGTGGCTCGATTCGATGCTGGAGAATTGATTACCCAGCGAGAGCTGGTCTCCAGGCAGGTGAGCGATGACCTCACAGAGCGAGCGGCAACATTTGGGCTTATCCTGGA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU010861EMU002781EMU034911
Ensembl | mouse accession no.

ENSMUSG00000038845

Ensembl | mouse accession no.

ENSMUSG00000034055

Ensembl | mouse accession no.

ENSMUSG00000029595

Ensembl | mouse accession no.

ENSMUSG00000040584

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

CTCATTTCCTCATCCCTTGGGTACAGAAACCAATTATCTTTGACTGCCGCTCTCGACCACGGAACGTACCAGTCATCACTGGTAGCAAAGACTTGCAGAACGTCAATATCACACTGCGAATCCTCTTCCGGCCGGTGGCCAGCCAGCTTCCTCGTATCTACACCAGCATTGGCGAGGACTATGATGAGCGGGTCCTGCCTTCTATCACCACAGAGATCCTCAAGTCGGTGGTGGCTCGATTCGATGCTGGAGAATTGATTACCCAGCGAGAGCTGGTCTCCAGGCAGGTGAGCGATGACCTCACAGAGCGAGCGGCAACATTTGGGCTTATCCTGGA

esiRNA cDNA target sequence

GGGGCTACTGTCAGAGAGCTTCTTAGTGAACTGTATGTCAAAGTTGGTGAAATTCGGCACTGGGGTCTGATCCGATATATCTCTGGGATCTTACGGAAGAAAGTGGAGGCACTTGATGAGGCCTGCACAGACCTTCTGTCCTACCAGAAACACCTGACAGTAGGACTTCCTCCAGAACCTCGAGAGAAGACCATCTCTGCGCCTCTACCGTACGAGGCACTCACTAAGCTGATAGATGAAGCCAGTGAAGGCGACATGAGCATCTCAACCCTTACACAGGAAATAATGGTCTATCTTGCCATGTATATGAGAACTCAGCCTGGCCTCTTTGCAGAAATGTTCAGACTTCGAATCGGTTTGATCATACAAGTTATGGCAACAGAACTAGCACACTCTCTTCGATGTTCAGCTGAGGAAGCC

esiRNA cDNA target sequence

AGTGCAAAACCAACCTCTCGGAGAAGTGCTTCTCACGGGAAGGCAAGCTATACTGTAAAAACGACTTTTTCAGGCGCTTTGGCACAAAGTGCGCCGGCTGCGCGCAAGGTATCTCTCCGAGCGACCTGGTACGGAAGGCCCGGAGCAAAGTCTTCCACCTCAACTGCTTCACCTGTATGGTGTGCAATAAGCAGCTATCCACCGGAGAGGAGCTCTACGTGATCGACGAGAACAAGTTTGTGTGCAAGGACGACTACTTAAGCTCCTCTAGCCTCAAGGAAGGAAGTCTCAACTCGGTGTCGTCCTGTACGGACCGCAGTTTGTCCCCGGACCTCCAGGATCCGTTACAGGACGACCCCAAAGAGACCGACAATTCGACCTCATCGGACAAGGAAACCGCTAACA

esiRNA cDNA target sequence

CAGCCCTGTTCTTGGACTGTCAGCTGGTATTTGGGCAAAGATATTGTCTTCATTTACTGATAAGGAACTCCATGCTTATGCAAAAGCTGGAGCAGTTGCTGAAGAAGTCTTAGCAGCCATCAGAACTGTGATTGCGTTTGGAGGACAAAAGAAGGAACTTGAAAGGTACAATAACAACTTGGAAGAAGCTAAAAGGCTGGGGATAAAGAAAGCTATCACGGCCAACATCTCCATGGGTGCAGCTTTTCTCCTTATCTATGCATCATATGCTCTGGCATTCTGGTATGGGACTTCCTTGGTCATCTCCAAAGAATACTCTATTGGACAAGTGCTCACTGTCTTCTTTTCCGTGTTAATTGGAGCATTCAGTGTTGGACAGGCATCTCCAAATATTGAAGCCTTCGCCAATGCACGAGGAGCAGCTTATGAAGTCTTCAAAATAATTGATAATAAGCCCAGTATAGACAGCTTCTCAAAGAGTGGGCAC

NCBI accession no.

NM_008831

NCBI accession no.

NM_173021

NCBI accession no.

NM_008499

NCBI accession no.

NM_011076

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satomi Miwa et al.
Nature communications, 5, 3837-3837 (2014-05-13)
Mitochondrial function is an important determinant of the ageing process; however, the mitochondrial properties that enable longevity are not well understood. Here we show that optimal assembly of mitochondrial complex I predicts longevity in mice. Using an unbiased high-coverage high-confidence

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service