Skip to Content
Merck
All Photos(1)

Key Documents

EMU189371

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Gtf3c3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCATAAGACAAGCTCCTTTGGCTTATGAGCCATTCTCTACACTTGCCATGATATACGAGGATCAAGGTGACATGGAGAAATCATTGCAGTTTGAGTTGATTGCTGCACATTTAAATCCCAGTGACACAGAAGAATGGGTTAGGCTGGCAGAAATGTCTCTGGAACAAGATAACATTAAGCAGGCTATTTTTTGCTATACAAAAGCTCTTAAATATGAACCTACTAATGTCCGTTACCTGTGGGAGAGATCAAGCCTTTATGAACAGATGGGTGATCATAAAATGGCCATGGATGGCTATAGGCGCATTTTAAATCTTTTGTCTCCATCTGATGGAGAACGTTTTATGCAGTTGGCTAGAGATATGGCAAAGAGTTACTATGAAGCCAATGACAGCGCTTCAGCTATAAACATAATTGAAGAAGCTTTCTCAAAACATCAAGGCTTAGTCTCCATGGAAGATGTTAACATAGCAGCTGAATTATATATTTCCAACAAGCAGTATGACAAAGC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU046801EMU065461EMU150481
product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

TCATAAGACAAGCTCCTTTGGCTTATGAGCCATTCTCTACACTTGCCATGATATACGAGGATCAAGGTGACATGGAGAAATCATTGCAGTTTGAGTTGATTGCTGCACATTTAAATCCCAGTGACACAGAAGAATGGGTTAGGCTGGCAGAAATGTCTCTGGAACAAGATAACATTAAGCAGGCTATTTTTTGCTATACAAAAGCTCTTAAATATGAACCTACTAATGTCCGTTACCTGTGGGAGAGATCAAGCCTTTATGAACAGATGGGTGATCATAAAATGGCCATGGATGGCTATAGGCGCATTTTAAATCTTTTGTCTCCATCTGATGGAGAACGTTTTATGCAGTTGGCTAGAGATATGGCAAAGAGTTACTATGAAGCCAATGACAGCGCTTCAGCTATAAACATAATTGAAGAAGCTTTCTCAAAACATCAAGGCTTAGTCTCCATGGAAGATGTTAACATAGCAGCTGAATTATATATTTCCAACAAGCAGTATGACAAAGC

esiRNA cDNA target sequence

GTTCCTGAACAGCCTTTGGAGGCTGCAGTCCAGACATGGAAGAAGGCCTGCACCAATCCCATAGATCAGAAGGTGGAAGAGGAGCTGAGGAAGCTCTTTGACATCCGTCCTGTTTGGTCACGAAATGCTGTCAAGTCTAACGTCAGTGTCCATCCTGACAAGCTTAAGATCTTGCTTCCCTATATGGCTTATTACATGATAACAGGCCCGTGGAGAAGCTTGTGGATTCGATTTGGCTATGACCCTCGAAAACACCCAGATGCCAAGATTTATCAAGTCCTTGATTTTCGAATCCGCTGTGGAATGAAATACGGTTATGGCTCCAGGGACATGCCTGTTAAAGCGAAGCGTAGCACGTACAACTACAGCCTGCCCATCACCGTCAAGAAGACATCCAACCAGCCTG

esiRNA cDNA target sequence

ATCATCCCGCAAAGTGAAAGTGGCAGGAAATCAGACATCAGCTGTGCTCCGAGGCCTGAAGAGCAACCTGGCCTATTACACAGCTGTCCGGGCTTACAACAGCGCAGGCGCTGGTCCCTTCAGCGCCACAGTTAATGCGACCACCAAGAAAACCCCTCCCAGTCAGCCACCAGGAAATGTTGTTTGGAATGCTACAGACACTAAAGTGCTACTTAATTGGGAACAAGTGAAAGCCATGGAGAATGAATCAGAAGTAACAGGCTACAAGGTTTTCTATAGGACTAGCAGTCAGAATAATGTGCACGTGCTGAACACAAATAAAACTTCAGCTGAACTGTTGCTGCCTATCAAAGAAGACTACATTATTGAGGTCAAGGCCACCACAGATGGAGGGGATGGCACCAGTAGTGAGCA

esiRNA cDNA target sequence

TCCACTCCCTTATTTCTGTTCCCTCCTTGTCCCTCAGGCAGAGTGACCTACAGCTTCCGGTAAAGACAGTGAGGGGCACTTAGACCAAGACTATATACATCAGGTTGGCCACAGCCTCTGTGCTTAGTGGAGTGGGCTGGGATTGTAGAAACTGCCCCAGGACTTGGCTTTGTGGGAATTTGTACACCCCACCTCCCTTTTTGACAAAATTATCTGAACTCAATATCTAGTGCATAAAGAATTTTTGTCAAATGGGTACATTTCGACTGCAGACTGACTCTCCTGCCTTAAACCAATCGGGTCCCCGCCTGGTCTCCTGACACATATGCTTTTTATGCTATGCCACATTGAAACATGTCCCGAGGGCTCACTGGTCCCCCACCATAGTGCAATATAGGACAGAGTCCCAGTCTTCATGAAGATAGGGAAGTTGGGAAATCCATATGAGATTATTACCAATGACACCGTTAAGATATATGATGCTTAAACCTTTTCAAAGCACTTTCATGT

Ensembl | mouse accession no.

ENSMUSG00000041303

Ensembl | mouse accession no.

ENSMUSG00000026816

Ensembl | mouse accession no.

ENSMUSG00000030075

Ensembl | mouse accession no.

ENSMUSG00000042734

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

NCBI accession no.

NM_001033194

NCBI accession no.

NM_148928

NCBI accession no.

NM_008779

NCBI accession no.

NM_001033149

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service