Skip to Content
Merck
All Photos(1)

Key Documents

EMU049681

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Rbm41

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Availability

Estimated to ship on17 February 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Availability

Estimated to ship on17 February 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCTAAGACGCTGAAGCAACCAATAGAATTTATCCCAGAAGATGAGATCCAGAGAAATCGTTTGTCAGAGGAGGAGATCCGAAACATTCCTATGTTTTCTTCATATAATCCAGGGGAACCAAACAAGGTGCTATATCTGAAGAACCTTAGTCCTCGGGTGAAAGAAAGAGACCTTATCTCATTATTCGCTCGATTCCAGGAGAAGAAAGGACCACCCATTCAGTTCCGAATGATGACTGGACGCATGAGGGGGCAAGCTTTCCTCACCTTTCCCAATAAAGACATAGCATGGCAAGCACTGCACCAAATTAATGGATATAAACTCTATGGGAAAATATTGGTGATAGAGTTTGCAAAGAGCAAAAAGCAACAGTCAGCTGTTCAGCGCTCTTCCCTTATAACATCTGCTCCTGATGGCAGAACAGAAA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU065661EMU051871EMU020431
Gene Information

mouse ... RBM41(237073), Rbm41(237073)

Gene Information

mouse ... RBM45(241490), Rbm45(241490)

Gene Information

mouse ... RBMS1(56878), Rbms1(56878)

Gene Information

mouse ... INSIG1(231070), Insig1(231070)

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Ensembl | mouse accession no.

ENSMUSG00000031433

Ensembl | mouse accession no.

ENSMUSG00000042369

Ensembl | mouse accession no.

ENSMUSG00000026970

Ensembl | mouse accession no.

ENSMUSG00000045294

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service