Skip to Content
Merck
All Photos(1)

Key Documents

EMU046811

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Atox1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGTGCCGCGTCAGTCATGCCGAAGCACGAGTTCTCCGTGGACATGACCTGTGAGGGCTGTGCTGAAGCCGTCTCCAGAGTCCTCAACAAGCTGGGAGGAGTGGAGTTCAACATTGACCTGCCCAACAAGAAGGTCTGCATCGACTCTGAGCACAGCTCAGACACCCTGCTGGCAACCCTCAACAAAACAGGAAAGGCTGTTTCCTACCTTGGCCCCAAGTAGCCAGGACCTGGGCGAGTCCTTCCGGATATAAACTGAAGAGGCAGGCTGTTGATCTGGTCTCCCCGGCAGATCTGGAACACCAACTGCTCAGTCCAGTCCAGCCCAGCCATGGAGTTCCTGCCCAGACAGGCCTTCCCCGCTGGCTCCCTGCAAGCTTCCATGTAATAAAGTCAAGCTGAGTTT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huawei Cai et al.
Oncology reports, 30(1), 269-275 (2013-04-30)
Copper is required for cell proliferation and tumor angiogenesis. Cellular copper metabolism is regulated by a network of copper transporters and chaperones. Antioxidant-1 (ATOX1) is a cytosolic copper chaperone important for intracellular copper transport, which plays a role in the
Arundhati Jana et al.
PloS one, 15(1), e0227916-e0227916 (2020-01-22)
Colorectal cancer remains a deadly cancer due to metastatic disease. To understand the molecular mechanisms of metastasis in colon cancer, we investigated whether the copper chaperone antioxidant-1 (Atox1) protein plays a role in this process. Recent findings indicate that Atox1
Gin-Fu Chen et al.
Scientific reports, 5, 14780-14780 (2015-10-07)
Copper (Cu), an essential micronutrient, plays a fundamental role in inflammation and angiogenesis; however, its precise mechanism remains undefined. Here we uncover a novel role of Cu transport protein Antioxidant-1 (Atox1), which is originally appreciated as a Cu chaperone and
Edward A Ratovitski
Current pharmaceutical biotechnology, 16(9), 832-850 (2015-06-20)
MicroRNAs, whose transcription is regulated by members of the tumor protein p53 family, modulate the expression of numerous metabolic enzymes, significantly altering tumor cell response to chemotherapeutic treatments. The role for ΔNp63α-regulated microRNAs in regulation of cell cycle arrest, apoptosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service