Skip to Content
Merck
All Photos(1)

Key Documents

EMU031691

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Casp8

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCAAATGAAATCCACGAGATTCTAGAAGGCTACCAAAGCGCAGACCACAAGAACAAAGACTGCTTCATCTGCTGTATCCTATCCCACGGTGACAAGGGTGTCGTCTATGGAACGGATGGGAAGGAGGCCTCCATCTATGACCTGACATCTTACTTCACTGGTTCAAAGTGCCCTTCCCTGTCTGGGAAACCCAAGATCTTTTTCATTCAGGCTTGCCAAGGAAGTAACTTCCAGAAAGGAGTGCCTGATGAGGCAGGCTTCGAGCAACAGAACCACACTTTAGAAGTGGATTCATCATCTCACAAGAACTATATTCCGGATGAGGCAGACTTTCTGCTGGGAATGGCTACGGTGAAGAACTGCGTTTCCTACCGAGATCCTGTGAATGGAACCTGGTATATTCAGTCACTTTGCCAGAGCCTGAGGGAAAGATGTCCTCAAGGAGATGACATTCTTAGCATCCTGACTGGCGT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Coriat et al.
Cell death & disease, 2, e191-e191 (2011-08-13)
Organotellurides are newly described redox-catalyst molecules with original pro-oxidative properties. We have investigated the in vitro and in vivo antitumoral effects of the organotelluride catalyst LAB027 in a mouse model of colon cancer and determined its profile of toxicity in
Meng Yu et al.
Molecular carcinogenesis, 53(7), 505-513 (2013-01-30)
Activation of telomerase is a key element in oncogenesis and resistance to apoptosis for many cancers. Some histone deacetylase inhibitors (HDACi) or chemotheraputic agents have been reported to downregulate the expression of human telomerase reverse transcriptase (hTERT). However, whether hTERT
René Weiss et al.
Antiviral research, 123, 93-104 (2015-09-15)
New anti-viral agents and strategies are urgently needed to fight rapidly mutating viruses, as vaccine programs cannot react fast enough to prevent pandemics. Recently, we have shown that interleukin-24 (IL-24) sensitizes tumor cells to toll-like receptor 3 (TLR3) mediated apoptosis.
Kei-Ichi Ishikawa et al.
PloS one, 9(4), e94645-e94645 (2014-04-12)
Mutations in p150glued cause hereditary motor neuropathy with vocal cord paralysis (HMN7B) and Perry syndrome (PS). Here we show that both overexpression of p150glued mutants and knockdown of endogenous p150glued induce apoptosis. Overexpression of a p150glued plasmid containing either a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service