Skip to Content
Merck
All Photos(1)

Key Documents

EMU030031

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Ddit3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GAATAACAGCCGGAACCTGAGGAGAGAGTGTTCCAGAAGGAAGTGCATCTTCATACACCACCACACCTGAAAGCAGAACCTGGTCCACGTGCAGTCATGGCAGCTGAGTCCCTGCCTTTCACCTTGGAGACGGTGTCCAGCTGGGAGCTGGAAGCCTGGTATGAGGATCTGCAGGAGGTCCTGTCCTCAGATGAAAATGGGGGCACCTATATCTCATCCCCAGGAAACGAAGAGGAAGAATCAAAAACCTTCACTACTCTTGACCCTGCGTCCCTAGCTTGGCTGACAGAGGAGCCAGGGCCAACAGAGGTCACACGCACATCCCAAAGCCCTCGCTCTCCAGATTCCAGTCAGAGTTCTATGGCCCAGGAGGAAGAGGAGGAAGAGCAAGGAAGAACTAGGAAACGGAAACAGAGTGGTCAGTGCCCAGCCCGGCCTGGGAAGCAACGCATGAAGGAGAAGGAGCAG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Aflaki et al.
Cell death & disease, 3, e280-e280 (2012-03-16)
Triacylglycerol (TG) accumulation caused by adipose triglyceride lipase (ATGL) deficiency or very low-density lipoprotein (VLDL) loading of wild-type (Wt) macrophages results in mitochondrial-mediated apoptosis. This phenotype is correlated to depletion of Ca(2+) from the endoplasmic reticulum (ER), an event known
Xin-Yu Zhang et al.
The international journal of biochemistry & cell biology, 68, 158-165 (2015-09-28)
Arsenic trioxide has been proven to trigger apoptosis in human hepatocellular carcinoma cells. Endoplasmic reticulum stress has been known to be involved in apoptosis through the induction of CCAAT/enhancer-binding protein homologous protein. However, it is unknown whether endoplasmic reticulum stress
Wen-Pin Cheng et al.
PloS one, 10(4), e0123235-e0123235 (2015-04-22)
The expression of TRB3 (tribbles 3), an apoptosis regulated gene, increases during endoplasmic reticulum (ER) stress. How mechanical stress affects the regulation of TRB3 in cardiomyocytes during apoptosis is not fully understood. An in vivo model of aorta-caval shunt in
Xiuhua Zhang et al.
BMC cancer, 15, 866-866 (2015-11-08)
Prostate cancer is the most commonly diagnosed malignancy among men. The Discovery of new agents for the treatment of prostate cancer is urgently needed. Compound WZ35, a novel analog of the natural product curcumin, exhibited good anti-prostate cancer activity, with
Lu Fan et al.
Toxicology and applied pharmacology, 278(1), 45-52 (2014-04-29)
Erlotinib, a popular drug for treating non-small cell lung cancer (NSCLC), causes diarrhea in approximately 55% of patients receiving this drug. In the present study, we found that erlotinib induced barrier dysfunction in rat small intestine epithelial cells (IEC-6) by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service