Skip to Content
Merck
All Photos(1)

Key Documents

EHU157031

Sigma-Aldrich

MISSION® esiRNA

targeting human NES

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGAGGCAGAGCTGAATCTGAGGGAGCAGGATGGCTTCACTGGGAAGGAGGAGGTGGTAGAGCAGGGAGAGCTGAATGCCACAGAGGAGGTCTGGATCCCAGGCGAGGGGCACCCAGAGAGCCCTGAGCCCAAAGAGCAGAGAGGCCTGGTTGAGGGAGCCAGTGTGAAGGGAGGGGCTGAGGGCCTCCAGGACCCTGAAGGGCAATCACAACAGGTGGGGGCCCCAGGCCTCCAGGCTCCCCAGGGGCTGCCAGAGGCGATAGAGCCCCTGGTGGAAGATGATGTGGCCCCAGGGGGTGACCAAGCCTCCCCAGAGGTCATGTTGGGGTCAGAGCCTGCCATGGGTGAGTCTGCTGCGGGAGCTGAGCCAGGCCCGGGGCAGGGGGTGGGAGGGCTGGGGGACCCAGGCCATCTGACCAGGGAAGAGGTGATGGAACCACCCCTGGAAGAGGAGAGTTTGGAGGCAAAGAGGGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Yang et al.
Kidney & blood pressure research, 43(2), 616-627 (2018-04-25)
Preeclampsia (PE) is a pregnancy-specific hypertensive disorder that is characterised by a high incidence of hypertension and proteinuria. Podocytes are involved in the formation of a split membrane, which is the last barrier preventing the leakage of protein into the
Yasuhiro Shinkai et al.
Nucleic acid therapeutics, 27(3), 168-175 (2017-03-30)
Herein we described the synthesis of siRNA-NES (nuclear export signal) peptide conjugates by solid phase fragment coupling and the application of them to silencing of bcr/abl chimeric gene in human chronic myelogenous leukemia cell line K562. Two types of siRNA-NES
Se Jeong Lee et al.
BMC cancer, 15, 1011-1011 (2015-12-26)
Glioblastoma multiforme (GBM) is characterized by extensive local invasion, which is in contrast with extremely rare systemic metastasis of GBM. Molecular mechanisms inhibiting systemic metastasis of GBM would be a novel therapeutic candidate for GBM in the brain. Patient-derived GBM
Kim Tardif et al.
American journal of physiology. Heart and circulatory physiology, 308(10), H1265-H1274 (2015-03-15)
Proliferation and hypertrophy of vascular smooth muscle cells represent hallmark features of vessel remodeling secondary to hypertension. The intermediate filament protein nestin was recently identified in vascular smooth muscle cells and in other cell types directly participated in proliferation. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service