Skip to Content
Merck
All Photos(1)

Key Documents

EHU156971

Sigma-Aldrich

MISSION® esiRNA

targeting human ERCC1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Estimated to ship on28 May 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Estimated to ship on28 May 2025


description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CGAATATGCCATCTCACAGCCTCTGGAAGGGGCTGGGGCCACGTGCCCCACAGGGTCAGAGCCCCTGGCAGGAGAGACGCCCAACCAGGCCCTGAAACCCGGGGCAAAATCCAACAGCATCATTGTGAGCCCTCGGCAGAGGGGCAATCCCGTACTGAAGTTCGTGCGCAATGTGCCCTGGGAATTTGGCGACGTAATTCCCGACTATGTGCTGGGCCAGAGCACCTGTGCCCTGTTCCTCAGCCTCCGCTACCACAACCTGCACCCAGACTACATCCATGGGCGGCTGCAGAGCCTGGGGAAGAACTTCGCCTTGCGGGTCCTGCTTGTCCAGGTGGATGTGAAAGATCCCCAGCAGGCCCTCAAGGAGCTGGCTAAGATGTGTATCCTGGCCGACTGCACATTGATCCTCGCCTGGAGCCCCGAGGAAGCTGGGCGGTACCTGGAGACCTACAAGGCCTATGAGCAGAAACCAGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jae Joon Han et al.
Cancer research and treatment : official journal of Korean Cancer Association, 46(1), 55-64 (2014-02-13)
The novel heat shock protein tumor necrosis factor receptor-associated protein 1 (TRAP1) is associated with multidrug resistance in colorectal cancer (CRC) cells in vitro. Excision repair cross-complementation group 1 (ERCC1) expression levels in tumor tissues also predict clinical outcomes in
Wei-Ping Lee et al.
Biochimica et biophysica acta, 1863(4), 917-928 (2017-01-16)
Gemcitabine and capecitabine are two effective anticancer agents against solid tumors. The pharmacological mechanisms have been known as incorporation into DNA and thereby inhibition of DNA synthesis. When used as metronomic chemotherapy, they may inhibit angiogenesis and induce immunity. In
Daniel P Feldmann et al.
Molecules (Basel, Switzerland), 25(8) (2020-04-30)
Platinum-based chemotherapy remains a mainstay treatment for the management of advanced non-small cell lung cancer. A key cellular factor that contributes to sensitivity to platinums is the 5'-3' structure-specific endonuclease excision repair cross-complementation group 1 (ERCC1)/ xeroderma pigmentosum group F
Gianmaria Liccardi et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 20(13), 3496-3506 (2014-05-02)
The epidermal growth factor receptor (EGFR) plays an important role in cellular response to chemotherapy and radiotherapy through modulation of DNA repair. EGFR activates DNA-dependent protein kinase (DNA-PK) stimulating repair of DNA strand breaks (SB) and interstrand crosslinks (ICL). We
Ying Wai Chan et al.
Nature cell biology, 20(1), 92-103 (2017-12-20)
The resolution of joint molecules that link recombining sister chromatids is essential for chromosome segregation. Here, we determine the fate of unresolved recombination intermediates arising in cells lacking two nucleases required for resolution (GEN1 -/- knockout cells depleted of MUS81).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service