Skip to Content
Merck
All Photos(1)

Key Documents

EHU155821

Sigma-Aldrich

MISSION® esiRNA

targeting human LITAF

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Estimated to ship on21 May 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Estimated to ship on21 May 2025


description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTCCTGCAACAAGATGATCGTGAGTCAGCTGTCCTATAACGCCGGTGCTCTGACCTGGCTGTCCTGCGGGAGCCTGTGCCTGCTGGGGTGCATAGCGGGCTGCTGCTTCATCCCCTTCTGCGTGGATGCCCTGCAGGACGTGGACCATTACTGTCCCAACTGCAGAGCTCTCCTGGGCACCTACAAGCGTTTGTAGGACTCAGCCAGACGTGGAGGGAGCCGGGTGCCGCAGGAAGTCCTTTCCACCTCTCATCCAGCTTCACGCCTGGTGGAGGTTCTGCCCTGGTGGTCTCACCTCTCCAGGGGGCCCACCTTCATGTCTTCTTTTGGGGGGAATACGTCGCAAAACTAACAAATCTCCAAACCCCAGAAATTGCTGCTTGGAGTCGTGCATAGGACTTGCAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Verdelli et al.
Laboratory investigation; a journal of technical methods and pathology, 97(12), 1488-1499 (2017-09-19)
Transcription factors active in embryonic parathyroid cells can be maintained in adult parathyroids and be involved in tumorigenesis. TBX1, the candidate gene of 22q11.2-DiGeorge syndrome, which includes congenital hypoparathyroidism, is involved in parathyroid embryogenesis. The study aimed to investigate expression
Changfeng Li et al.
International journal of molecular medicine, 45(3), 956-966 (2020-01-11)
Colorectal cancer is a commonly diagnosed gastrointestinal malignancy worldwide with a high mortality rate. Accumulating evidence has indicated that the expression of a number of microRNAs (miRNAs) is associated with the development of colorectal cancer. However, the precise molecular mechanism

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service