Skip to Content
Merck
All Photos(1)

Documents

EHU119911

Sigma-Aldrich

MISSION® esiRNA

targeting human DAPK3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCATGCTGCTGGACAAGAACGTGCCCAACCCACGAATCAAGCTCATCGACTTCGGCATCGCGCACAAGATCGAGGCGGGGAACGAGTTCAAGAACATCTTCGGCACCCCGGAGTTTGTGGCCCCAGAGATTGTGAACTATGAGCCGCTGGGCCTGGAGGCGGACATGTGGAGCATCGGTGTCATCACCTATATCCTCCTGAGCGGTGCATCCCCGTTCCTGGGCGAGACCAAGCAGGAGACGCTCACCAACATCTCAGCCGTGAACTACGACTTCGACGAGGAGTACTTCAGCAACACCAGCGAGCTGGCCAAGGACTTCATTCGCCGGCTGCTCGTCAAAGATCCCAAGCGGAGAATGACCATTGCCCAGAGCCTGGAACATTCCTGGATTAAGGCGATCCGGCGGCGGAACGTGCGTGGTGAGGACAGCGGCCGCAAGCCCGAGCGGCGGCGCCTGAAGACCACGCGTCTGAAGGAGTACACCATCAAGTCGCACTCCAGCTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ke-Xue Li et al.
European journal of pharmacology, 852, 90-98 (2019-03-10)
Vascular calcification (VC) is a critical feature of chronic kidney disease (CKD), diabetes, hypertension, and atherosclerosis. Death-associated protein kinase 3 (DAPK3) is involved in vascular remodeling in hypertension. However, it remains to be clarified whether DAPK3 controls vascular smooth muscle
Jing-Ti Deng et al.
PloS one, 14(12), e0226406-e0226406 (2019-12-14)
Myosin regulatory light chain (LC20) phosphorylation plays an important role in vascular smooth muscle contraction and cell migration. Ca2+/calmodulin-dependent myosin light chain kinase (MLCK) phosphorylates LC20 (its only known substrate) exclusively at S19. Rho-associated kinase (ROCK) and zipper-interacting protein kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service