Skip to Content
Merck
All Photos(2)

Key Documents

EHU114671

Sigma-Aldrich

MISSION® esiRNA

targeting human HSP90AA1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGTGACATCTCCATGCTGTATTGTCACAAGCACATATGGCTGGACAGCAAACATGGAGAGAATCATGAAAGCTCAAGCCCTAAGAGACAACTCAACAATGGGTTACATGGCAGCAAAGAAACACCTGGAGATAAACCCTGACCATTCCATTATTGAGACCTTAAGGCAAAAGGCAGAGGCTGATAAGAACGACAAGTCTGTGAAGGATCTGGTCATCTTGCTTTATGAAACTGCGCTCCTGTCTTCTGGCTTCAGTCTGGAAGATCCCCAGACACATGCTAACAGGATCTACAGGATGATCAAACTTGGTCTGGGTATTGATGAAGATGACCCTACTGCTGATGATACCAGTGCTGCTGTAACTGAAGAAATGCCACCCCTTGAAGGAGATGACGACACATCACGCATGGAAGAAGTAGACTAATCTCTGGCTGAGGGATGACT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Xiao et al.
Journal of experimental & clinical cancer research : CR, 37(1), 201-201 (2018-08-30)
Osteosarcoma is the most common primary bone tumor in children and adolescents. Unfortunately, osteosarcoma treatments often fail due to the development of chemoresistance, of which the underlying molecular mechanisms still remain unclear. In this study, we demonstrated that HSP90AA1 gene
Robert O'Brien et al.
The Journal of biological chemistry, 290(31), 19287-19306 (2015-05-31)
The cascade of events that lead to cognitive decline, motor deficits, and psychiatric symptoms in patients with Huntington disease (HD) is triggered by a polyglutamine expansion in the N-terminal region of the huntingtin (HTT) protein. A significant mechanism in HD

Articles

Quantitative and qualitative western blotting to validate knockdown by esiRNA.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service