Skip to Content
Merck
All Photos(1)

Documents

EHU114221

Sigma-Aldrich

MISSION® esiRNA

targeting human YY1AP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAGACCAAACAGCTGCCAGTCCTAGGAAAATGCTGTGAAGAGATCCAGCCACATCAGTGGAAGCCACCTATAGAGAGAGAAGAACACCGGCTCCCATTCTGGTTAAAGGCCAGTCTGCCATCCATCCAGGAAGAACTGCGGCACATGGCTGATGGTGCTAGAGAGGTAGGAAATATGACTGGAACCACTGAGATCAACTCAGATCAAGGCCTAGAAAAAGACAACTCAGAGTTGGGGAGTGAAACTCGGTACCCACTGCTATTGCCTAAGGGTGTAGTCCTGAAACTGAAGCCAGTTGCCGACCGTTTCCCCAAGAAGGCTTGGAGACAGAAGCGTTCATCAGTCCTGAAACCCCTCCTTATCCAACCCAGCCCCTCTCTCCAGCCCAGCTTCAACCCTGGGAAAACAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C He et al.
Oncogene, 34(50), 6040-6054 (2015-03-24)
Mechanisms underlying ovarian cancer initiation and progression are unclear. Herein, we report that the Yes-associated protein (YAP), a major effector of the Hippo tumor suppressor pathway, interacts with ERBB signaling pathways to regulate the initiation and progression of ovarian cancer.
David Fu et al.
Endocrine-related cancer, 21(2), 297-310 (2014-01-07)
The Hippo signaling pathway has been implicated as a conserved regulator of organ size in both Drosophila and mammals. Yes-associated protein (YAP), the central component of the Hippo signaling cascade, functions as an oncogene in several malignancies. Ovarian granulosa cell
Hyun-Jung Choi et al.
Nature communications, 6, 6943-6943 (2015-05-13)
Angiogenesis is regulated by the dynamic interaction between endothelial cells (ECs). Hippo-Yes-associated protein (YAP) signalling has emerged as a key pathway that controls organ size and tissue growth by mediating cell contact inhibition. However, the role of YAP in EC
Upal Basu-Roy et al.
Nature communications, 6, 6411-6411 (2015-04-04)
The repressive Hippo pathway has a profound tumour suppressive role in cancer by restraining the growth-promoting function of the transcriptional coactivator, YAP. We previously showed that the stem cell transcription factor Sox2 maintains cancer stem cells (CSCs) in osteosarcomas. We
Ute Schütte et al.
Translational oncology, 7(2), 309-321 (2014-06-11)
Recent work has identified dysfunctional Hippo signaling to be involved in maintenance and progression of various human cancers, although data on clear cell renal cell carcinoma (ccRCC) have been limited. Here, we provide evidence implicating aberrant Hippo signaling in ccRCC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service