Skip to Content
Merck
All Photos(1)

Key Documents

EHU109691

Sigma-Aldrich

MISSION® esiRNA

targeting human HSPA5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTATGAAGCCCGTCCAGAAAGTGTTGGAAGATTCTGATTTGAAGAAGTCTGATATTGATGAAATTGTTCTTGTTGGTGGCTCGACTCGAATTCCAAAGATTCAGCAACTGGTTAAAGAGTTCTTCAATGGCAAGGAACCATCCCGTGGCATAAACCCAGATGAAGCTGTAGCGTATGGTGCTGCTGTCCAGGCTGGTGTGCTCTCTGGTGATCAAGATACAGGTGACCTGGTACTGCTTGATGTATGTCCCCTTACACTTGGTATTGAAACTGTGGGAGGTGTCATGACCAAACTGATTCCAAGGAACACAGTGGTGCCTACCAAGAAGTCTCAGATCTTTTCTACAGCTTCTGATAATCAACCAACTGTTACAATCAAGGTCTATGAAGGTGAAAGACCCCTGACAAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patricia Dauer et al.
Molecular oncology, 12(9), 1498-1512 (2018-05-09)
Chemoresistance is a major therapeutic challenge that plays a role in the poor statistical outcomes in pancreatic cancer. Unfolded protein response (UPR) is one of the homeostasis mechanisms in cancer cells that have been correlated with chemoresistance in a number
Hiroshi Kokubun et al.
International journal of molecular sciences, 21(2) (2020-01-16)
Preclinical studies have shown that exposure of the developing brain to inhalational anesthetics can cause neurotoxicity. However, other studies have claimed that anesthetics can exert neuroprotective effects. We investigated the mechanisms associated with the neurotoxic and neuroprotective effects exerted by
Kyung-Won Park et al.
Scientific reports, 7, 44723-44723 (2017-03-24)
Prion diseases are fatal neurodegenerative disorders affecting several mammalian species, characterized by the accumulation of the misfolded form of the prion protein, which is followed by the induction of endoplasmic reticulum (ER) stress and the activation of the unfolded protein
Haopeng Yang et al.
Oncotarget, 7(50), 83641-83656 (2016-11-16)
Pancreatic cancer is a highly aggressive malignancy, which is intrinsically resistant to current chemotherapies. Herein, we investigate whether bisdemethoxycurcumin (BDMC), a derivative of curcumin, potentiates gemcitabine in human pancreatic cancer cells. The result suggests that BDMC sensitizes gemcitabine by inducing
Xiao-Qiang Wang et al.
The Journal of steroid biochemistry and molecular biology, 172, 188-197 (2017-06-25)
17beta-hydroxysteroid dehydrogenase type 7 (17β-HSD7) promotes breast cancer cell growth via dual-catalytic activity by modulating estradiol and DHT. Here, we clarified the expression pattern of 17β-HSD7 in postmenopausal luminal A type breast cancer with The Cancer Genome Atlas (TCGA) cohort.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service