Skip to Content
Merck
All Photos(2)

Key Documents

EHU106441

Sigma-Aldrich

MISSION® esiRNA

targeting human PTEN

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACCGCCAAATTTAATTGCAGAGTTGCACAATATCCTTTTGAAGACCATAACCCACCACAGCTAGAACTTATCAAACCCTTTTGTGAAGATCTTGACCAATGGCTAAGTGAAGATGACAATCATGTTGCAGCAATTCACTGTAAAGCTGGAAAGGGACGAACTGGTGTAATGATATGTGCATATTTATTACATCGGGGCAAATTTTTAAAGGCACAAGAGGCCCTAGATTTCTATGGGGAAGTAAGGACCAGAGACAAAAAGGGAGTAACTATTCCCAGTCAGAGGCGCTATGTGTATTATTATAGCTACCTGTTAAAGAATCATCTGGATTATAGACCAGTGGCACTGTTGTTTCACAAGATGATGTTTGAAACTATTCCAATGTTCAGTGGCGGAACTTGCAATCCTCAGTTTGTGGTCTGCCAGCTAAAGGTGAAGATATATTCCTCCAATTCAGGACCCACACGACGGGAAGACAAGTTCATGTACTTTGAGTTCCCTCAGCCGTTACCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Dou et al.
Frontiers in oncology, 11, 614035-614035 (2021-03-27)
microRNAs (miRNAs) are of great significance in cancer treatment, which may have a desirable result on the regulation of tumorigenesis, progression, recurrence, and chemo-resistance of ovarian cancer. However, the research on the further potential application of miR-4461 in ovarian cancer
Hu Gao et al.
Reproduction (Cambridge, England) (2019-11-23)
Sertoli cells are indispensable for normal spermatogenesis and increasing evidence has shown that microRNAs (miRNAs) participate in the regulation of Sertoli cell growth. However, the functions and regulatory mechanisms of miRNAs in Sertoli cells of domestic animals have not been
Aram Ghalali et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 127, 110112-110112 (2020-04-16)
Akt kinase regulates several cellular processes, among them growth, proliferation and survival, and has been correlated to neoplastic disease. We report here crosstalk between several Akt regulatory phosphatases that controls the level of the activated form (phosphorylated) of Akt and
Ye Wang et al.
OncoTargets and therapy, 13, 1909-1919 (2020-03-19)
Berberine (BBR), a traditional Chinese medicine, has been shown effects on inhibiting cancer development. Autophagy-mediated resistance plays an important role in cancer progression; therefore, regulation of autophagy is a novel therapeutic strategy for cancer treatment. However, effects of BBR on
Xiaomei Zou et al.
Frontiers in pharmacology, 11, 880-880 (2020-06-26)
Neuronal insulin resistance is implicated in neurodegenerative diseases. Icariin has been reported to improve insulin resistance in skeletal muscle cells and to restore impaired hypothalamic insulin signaling in the rats with chronic unpredictable mild stress. In addition, icariin can exert

Articles

Quantitative and qualitative western blotting to validate knockdown by esiRNA.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service