Skip to Content
Merck
All Photos(1)

Key Documents

EHU106111

Sigma-Aldrich

MISSION® esiRNA

targeting human KRT17

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CGCACCAAGTTTGAGACAGAGCAGGCCCTGCGCCTGAGTGTGGAGGCCGACATCAATGGCCTGCGCAGGGTGCTGGATGAGCTGACCCTGGCCAGAGCCGACCTGGAGATGCAGATTGAGAACCTCAAGGAGGAGCTGGCCTACCTGAAGAAGAACCACGAGGAGGAGATGAACGCCCTGCGAGGCCAGGTGGGTGGTGAGATCAATGTGGAGATGGACGCTGCCCCAGGCGTGGACCTGAGCCGCATCCTCAACGAGATGCGTGACCAGTATGAGAAGATGGCAGAGAAGAACCGCAAGGATGCCGAGGATTGGTTCTTCAGCAAGACAGAGGAACTGAACCGCGAGGTGGCCACCAACAGTGAGCTGGTGCAGAGTGGCAAGAGTGAGATCTCGGAGCTCCGGCGCACCATGCAGGCCTTGGAGATAGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chun-Ying Xiao et al.
Chinese medical journal, 133(24), 2910-2918 (2020-11-26)
Psoriasis is a common chronic inflammatory skin disease with 2% to 3% prevalence worldwide and a heavy social-psychological burden for patients and their families. As the exact pathogenesis of psoriasis is still unknown, the current treatment is far from satisfactory.
Daisuke Ujiie et al.
Carcinogenesis, 41(5), 591-599 (2019-11-23)
Adjuvant chemotherapy is considered for patients with stage II colorectal cancer (CRC) characterized by poor prognostic clinicopathological features; however, current stratification algorithms remain inadequate for identifying high-risk patients. To develop prognostic assays, we conducted a step-wise screening and validation strategy
Mihaela Chivu-Economescu et al.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association, 20(6), 948-959 (2017-03-17)
Keratin 17 (KRT17) was shown to be an important molecular marker for predicting the carcinogenesis, progression, and prognosis of various cancer types. Our previous studies identified KRT17 as a possible biomarker for gastric cancer by gene microarray, with an elevated
Hui Liu et al.
Gene, 563(1), 35-40 (2015-03-10)
Hereditary protein C deficiency (PCD) is an autosomal inherited disorder associated with high risk for venous thromboembolism (VTE). This study aimed to explore the functional consequences of two missense mutations, p.Asp297His and p.Val420Ile, responsible for type I/II PCD and recurrent
Jason D Arroyo et al.
Nucleic acids research, 42(9), 6064-6077 (2014-03-07)
Unlike short interfering RNAs (siRNAs), which are commonly designed to repress a single messenger RNA (mRNA) target through perfect base pairing, microRNAs (miRNAs) are endogenous small RNAs that have evolved to concurrently repress multiple mRNA targets through imperfect complementarity. MicroRNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service