Skip to Content
Merck
All Photos(1)

Key Documents

EHU106031

Sigma-Aldrich

MISSION® esiRNA

targeting human CDC20

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTACAGCCAAAAGGCCACTCCTGGCTCCAGCCGGAAGACCTGCCGTTACATTCCTTCCCTGCCAGACCGTATCCTGGATGCGCCTGAAATCCGAAATGACTATTACCTGAACCTTGTGGATTGGAGTTCTGGGAATGTACTGGCCGTGGCACTGGACAACAGTGTGTACCTGTGGAGTGCAAGCTCTGGTGACATCCTGCAGCTTTTGCAAATGGAGCAGCCTGGGGAATATATATCCTCTGTGGCCTGGATCAAAGAGGGCAACTACTTGGCTGTGGGCACCAGCAGTGCTGAGGTGCAGCTATGGGATGTGCAGCAGCAGAAACGGCTTCGAAATATGACCAGTCACTCTGCCCGAGTGGGCTCCCTAAGCTGGAACAGCTATATCCTGTCCAGTGGTTCACGTTCTGGCCACATCCACCACCATGATGT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuan Gao et al.
Aging, 13(2), 2668-2680 (2021-01-08)
The molecular mechanism of osteosarcoma (OS) pathogenesis is poorly understood. The Notch signaling pathway has been shown to be critically involved in tumorigenesis, including OS. Therefore, we explored the molecular mechanism by which the Notch-1 signaling pathway is involved in
Shujie Cheng et al.
International journal of oncology, 54(6), 2250-2256 (2019-05-14)
Aberrant expression of cell division cycle 20 (CDC20) is associated with malignant progression and poor prognosis in various types of cancer. The development of specific CDC20 inhibitors may be a novel strategy for the treatment of cancer with elevated expression of
Jia Li et al.
International journal of oncology, 45(4), 1547-1555 (2014-07-30)
Cell division cycle 20 (CDC20) encodes a regulatory protein interacting with the anaphase-promoting complex/cyclosome (APC/C) in the cell cycle and plays important roles in tumorigenesis and progression of multiple tumors. The present study aimed to investigate the clinical significance of
N Bah et al.
Cell death & disease, 5, e1291-e1291 (2014-06-13)
Antimitotic agents such as microtubule inhibitors (paclitaxel) are widely used in cancer therapy while new agents blocking mitosis onset are currently in development. All these agents impose a prolonged mitotic arrest in cancer cells that relies on sustained activation of
Karine Boulay et al.
Nucleic acids research, 42(12), 7867-7883 (2014-06-08)
Staufen1 (Stau1) is a ribonucleic acid (RNA)-binding protein involved in the post-transcriptional regulation of gene expression. Recent studies indicate that Stau1-bound messenger RNAs (mRNAs) mainly code for proteins involved in transcription and cell cycle control. Consistently, we report here that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service