Skip to Content
Merck
All Photos(1)

Key Documents

EHU102911

Sigma-Aldrich

MISSION® esiRNA

targeting human RACK1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGAGTGTGGCCTTCTCCTCTGACAACCGGCAGATTGTCTCTGGATCTCGAGATAAAACCATCAAGCTATGGAATACCCTGGGTGTGTGCAAATACACTGTCCAGGATGAGAGCCACTCAGAGTGGGTGTCTTGTGTCCGCTTCTCGCCCAACAGCAGCAACCCTATCATCGTCTCCTGTGGCTGGGACAAGCTGGTCAAGGTATGGAACCTGGCTAACTGCAAGCTGAAGACCAACCACATTGGCCACACAGGCTATCTGAACACGGTGACTGTCTCTCCAGATGGATCCCTCTGTGCTTCTGGAGGCAAGGATGGCCAGGCCATGTTATGGGATCTCAACGAAGGCAAACACCTTTACACGCTAGATGGTGGGGACATCATCAACGCCCTGTGCTTCAGCCCTAACCGCTACTGGCTGTGTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Hu et al.
Cancer letters, 450, 144-154 (2019-03-09)
Receptor of activated protein kinase C 1 (RACK1) is downregulated in gastric cancer and is involved in modulating NF-κB signaling pathway activity. However, the underlying molecular mechanisms regulating RACK1 expression are unclear. In this study, we demonstrated that downregulated expression
Lei Zhang et al.
OncoTargets and therapy, 12, 1007-1020 (2019-02-19)
The expression and function of the Receptor for Activated C Kinase 1 (RACK1) in cancer growth and metastasis are confused in different cancers, especially in pancreatic ductal adenocarcinoma (PDAC). One-hundred and eighty-two PDAC tissue specimens (95 males and 87 females)
Wenting He et al.
Journal of Alzheimer's disease : JAD, 75(2), 451-460 (2020-04-07)
Accumulation of amyloid-β (Aβ) peptides, generated from amyloid-β precursor protein (AβPP) amyloidogenic processing, is one of the most salient disease hallmarks of Alzheimer's disease (AD). Nicotine is able to promote α-secretase-mediated AβPP nonamyloidogenic processing and increase the release of sAβPPα
Zhao-Fei Dong et al.
Molecular neurobiology, 50(2), 438-448 (2014-01-18)
Voltage-gated sodium channel α subunit type I (Nav1.1, encoded by SCN1A gene) plays a critical role in the initiation of action potential in the central nervous system. Downregulated expression of SCN1A is believed to be associated with epilepsy. Here, we
Vasilios A Morikis et al.
Journal of leukocyte biology, 108(6), 1815-1828 (2020-06-13)
Recruitment of leukocytes to sites of acute inflammation is guided by spatial and temporal cues that ensure appropriate cell numbers infiltrate the tissue at precise locations to protect it from infection and initiate repair. On inflamed endothelium, neutrophil rolling via

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service