Skip to Content
Merck
All Photos(1)

Key Documents

EHU100561

Sigma-Aldrich

MISSION® esiRNA

targeting human CLEC7A

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Estimated to ship on01 June 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Estimated to ship on01 June 2025


description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGCACCCAGCCTAGAATCTTGTATAATATGTAATTGTAGGGAAACTGCTCTCATAGGAAAGTTTTCTGCTTTTTAAATACAAAAATACATAAAAATACATAAAATCTGATGATGAATATAAAAAAGTAACCAACCTCATTGGAACAAGTATTAACATTTTGGAATATGTTTTATTAGTTTTGTGATGTACTGTTTTACAATTTTTACCATTTTTTTCAGTAATTACTGTAAAATGGTATTATTGGAATGAAACTATATTTCCTCATGTGCTGATTTGTCTTATTTTTTTCATACTTTCCCACTGGTGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther Weiss et al.
Frontiers in cellular and infection microbiology, 8, 288-288 (2018-09-05)
Invasive aspergillosis (IA) is an infectious disease caused by the fungal pathogen Aspergillus fumigatus that mainly affects immunocompromised hosts. To investigate immune cell cross-talk during infection with A. fumigatus, we co-cultured natural killer (NK) cells and dendritic cells (DC) after
Cheng-Ye Che et al.
International journal of ophthalmology, 11(6), 905-909 (2018-07-07)
To investigate the regulation of lipoxygenase (LOX)-1 and Dectin-1 on interleukin-10 (IL-10) production in mice with Aspergillus fumigatus (A. fumigatus) keratitis. The corneas of C57BL/6 mice were pretreated with LOX-1 inhibitor Poly(I) or Dectin-1 siRNA separately before the infection of
Nobuaki Fujiwara et al.
Cancer gene therapy, 26(1-2), 32-40 (2018-07-05)
Antisense oligonucleotides (AS-ODNs) hybridize with specific mRNAs, resulting in interference with the splicing mechanism or the regulation of protein translation. We previously demonstrated that the β-glucan schizophyllan (SPG) can form a complex with AS-ODNs with attached dA40 (AS-ODNs/SPG), and this
Kelan Yuan et al.
International immunopharmacology, 52, 168-175 (2017-09-20)
To investigate the role of phosphorylated JNK in Dectin-1-induced IL-1β production and the role of Dectin-1 in apoptosis in mouse Aspergillus fumigatus (A. fumigatus) keratitis. Mice corneas were pretreated with Dectin-1 siRNA or SP600125 (the inhibitor of JNK) before A.
Xia Hua et al.
PloS one, 10(6), e0128039-e0128039 (2015-06-04)
Fungal infections of the cornea can be sight-threatening and have a worse prognosis than other types of microbial corneal infections. Peptidoglycan recognition proteins (PGLYRP), which are expressed on the ocular surface, play an important role in the immune response against

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service