Skip to Content
Merck
All Photos(1)

Key Documents

EHU094591

Sigma-Aldrich

MISSION® esiRNA

targeting human MYB

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Estimated to ship on20 May 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Estimated to ship on20 May 2025


description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGGCAGTAGAGCTTGGACAGAAAGAAAAGAAACTTGGTGTTAGGTAATTGACTATGCACTAGTATTTCAGACTTTTTAATTTTATATATATATACATTTTTTTTCCTTCTGCAATACATTTGAAAACTTGTTTGGGAGACTCTGCATTTTTTATTGTGGTTTTTTTGTTATTGTTGGTTTATACAAGCATGCGTTGCACTTCTTTTTTGGGAGATGTGTGTTGTTGATGTTCTATGTTTTGTTTTGAGTGTAGCCTGACTGTTTTATAATTTGGGAGTTCTGCATTTGATCCGCATCCCCTGTGGTTTCTAAGTGTATGGTCTCAGAACTGTTGCATGGATCCTGTGTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Wang et al.
Cancer letters, 385, 234-242 (2016-10-25)
The oncoprotein Yes-associated protein (YAP) in Hippo pathway plays crucial roles in the development of cancer. However, the mechanism of YAP regulation in cancer remains poorly understood. Here, we supposed that the oncoprotein hepatitis B X-interacting protein (HBXIP) might be
Tian Lan et al.
Cancer research, 79(13), 3220-3234 (2019-05-19)
Understanding the roles of noncoding RNAs (ncRNA) in tumorigenesis and metastasis would establish novel avenues to identify diagnostic and therapeutic targets. Here, we aimed to identify hepatocellular carcinoma (HCC)-specific ncRNA and to investigate their roles in hepatocarcinogenesis and metastasis. RNA-seq
Andrej Gorbatenko et al.
Scientific reports, 9(1), 3352-3352 (2019-03-06)
The HER2 oncogene and its truncated form p95HER2 play central roles in breast cancer. Here, we show that although HER2 and p95HER2 generally elicit qualitatively similar changes in miRNA profile in MCF-7 breast cancer cells, a subset of changes are
Xuefeng Li et al.
Nature microbiology, 1(10), 16132-16132 (2016-09-28)
MicroRNAs (miRNAs) play critical roles in various biological processes, including cell proliferation, development and host defence. However, the molecular mechanism for miRNAs in regulating bacterial-induced inflammation remains largely unclear. Here, we report that miR-301b augments pro-inflammatory response during pulmonary infection
P Basova et al.
Oncogene, 33(39), 4735-4745 (2013-10-15)
PU.1 downregulation within hematopoietic stem and progenitor cells (HSPCs) is the primary mechanism for the development of acute myeloid leukemia (AML) in mice with homozygous deletion of the upstream regulatory element (URE) of PU.1 gene. p53 is a well-known tumor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service