Skip to Content
Merck
All Photos(1)

Key Documents

EHU093581

Sigma-Aldrich

MISSION® esiRNA

targeting human CTNND1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTGGCTGAAGTTGAACATGGGAGCTTATTGCTAATTTAGAGATAGGAAACTGAAGCATAAAGAATTAATGACTTACTTTAATTACTGGAATTCTTCTGCAACATTTGACAAAACTAACCTTGAATAAGGCCCACTGTAATACGTAGCTCTCTTAAATATAACACTTAGGACTAGAAGATTAGAAACTACCAATCCCAACTACGTAATAGGAAAATGTAGGATCAAAAGGCCCATGTATATAAGTACTGACCACTGGGCCATAATGTTGCTTCTCAGGCTATATGCAGTCCTTTAGTCAGAAGTCAATAGGCCTATTTATTAATATTTTACAGACCATATTACCTGGATTACCAGGGACTATCTTTGCTGC

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alisha M Mendonsa et al.
PloS one, 15(6), e0235337-e0235337 (2020-06-27)
p120-catenin is considered to be a tumor suppressor because it stabilizes E-cadherin levels at the cell surface. p120-catenin phosphorylation is increased in several types of cancer, but the role of phosphorylation in cancer is unknown. The phosphorylation state of p120-catenin
Ningjia Cao et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 106, 483-490 (2018-07-11)
MicroRNAs (miRNAs) are solid factors involved in the initiation and progression of hepatocellular carcinoma (HCC). Recently, miR-298 is recognized as a cancer-associated miRNA in breast, gastric and ovarian cancer. However, the functional role of miR-298 and its underlying mechanism are
Xiao-Hui Gao et al.
Scientific reports, 10(1), 44-44 (2020-01-09)
Low miR-96-5p expression is characteristic of many cancers but its role in breast cancer (BCa) remains poorly defined. Here, the role of miR-96-5p in BC development was assessed. We demonstrate that exogenously expressing miR-96-5p inhibits the proliferative, migratory and invasive
Rong Wang et al.
Molecular carcinogenesis, 56(7), 1733-1742 (2017-02-22)
The heterocyclic amine 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) targets multiple organs for tumorigenesis in the rat, including the colon and the skin. PhIP-induced skin tumors were subjected to mutation screening, which identified genetic changes in Hras (7/40, 17.5%) and Tp53 (2/40, 5%), but
Josué Curto et al.
Molecular oncology, 12(5), 611-629 (2018-02-22)
Canonical and noncanonical Wnt pathways share some common elements but differ in the responses they evoke. Similar to Wnt ligands acting through the canonical pathway, Wnts that activate the noncanonical signaling, such as Wnt5a, promote Disheveled (Dvl) phosphorylation and its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service