Skip to Content
Merck
All Photos(1)

Key Documents

EHU092951

Sigma-Aldrich

MISSION® esiRNA

targeting human CREB1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGAGTGCCAAGGATTGAAGAAGAGAAGTCTGAAGAGGAGACTTCAGCACCTGCCATCACCACTGTAACGGTGCCAACTCCAATTTACCAAACTAGCAGTGGACAGTATATTGCCATTACCCAGGGAGGAGCAATACAGCTGGCTAACAATGGTACCGATGGGGTACAGGGCCTGCAAACATTAACCATGACCAATGCAGCAGCCACTCAGCCGGGTACTACCATTCTACAGTATGCACAGACCACTGATGGACAGCAGATCTTAGTGCCCAGCAACCAAGTTGTTGTTCAAGCTGCCTCTGGAGACGTACAAACATACCAGATTCGCACAGCACCCACTAGCACTATTGCCCCTGGAGTTGTTATGGCATCCTCCCCAGCACTTCCTACACAGCCTGCTGAAGAAGCAGCACGAAAGAGAGAGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Supriya Srinivasan et al.
Cancer research, 78(21), 6146-6158 (2018-09-21)
Although smoking is a significant risk factor for pancreatic ductal adenocarcinoma (PDAC), the molecular mechanisms underlying PDAC development and progression in smokers are still unclear. Here, we show the role of cyclic AMP response element-binding protein (CREB) in the pathogenesis
Hui-Xia Geng et al.
Neurochemical research, 42(10), 2841-2849 (2017-05-17)
Neuronal apoptosis mediated by the mitochondrial apoptosis pathway is an important pathological process in cerebral ischemia-reperfusion injury. 14,15-EET, an intermediate metabolite of arachidonic acid, can promote cell survival during ischemia/reperfusion. However, whether the mitochondrial apoptotic pathway is involved this survival
L Yuan et al.
Neuropathology and applied neurobiology, 42(7), 607-620 (2015-11-04)
14,15-Epoxyeicosatrienoic acid (14,15-EET) is abundantly expressed in brain and exerts protective effects against ischaemia. 14,15-EET is hydrolysed by soluble epoxide hydrolase (sEH). sEH-/- mice show a higher level of 14,15-EET in the brain. Astrocytes play a pivotal role in neuronal
Tao Xiao et al.
Stem cells translational medicine, 8(2), 148-161 (2018-11-15)
Fibrous dysplasia (FD) is a disease of postnatal skeletal stem cells caused by activating mutations of guanine nucleotide-binding protein alpha-stimulating activity polypeptide (GNAS). FD is characterized by high proliferation and osteogenesis disorder of bone marrow stromal cells (BMSCs), resulting in
Monika Sharma et al.
Journal of neuroimmunology, 332, 37-48 (2019-04-02)
Fundamentally, microglia have two activation states, a pro-inflammatory neurotoxic (M1) and an anti-inflammatory neuroprotective (M2) phenotype, and their conversion from M1-like to M2-like microglia may provide therapeutic benefits to prevent neuronal loss in neurodegenerative diseases such as Parkinson's disease (PD).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service