Skip to Content
Merck
All Photos(1)

Key Documents

EHU090991

Sigma-Aldrich

MISSION® esiRNA

targeting human PAX6

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATGAGGCTCAAATGCGACTTCAGCTGAAGCGGAAGCTGCAAAGAAATAGAACATCCTTTACCCAAGAGCAAATTGAGGCCCTGGAGAAAGAGTTTGAGAGAACCCATTATCCAGATGTGTTTGCCCGAGAAAGACTAGCAGCCAAAATAGATCTACCTGAAGCAAGAATACAGGTATGGTTTTCTAATCGAAGGGCCAAATGGAGAAGAGAAGAAAAACTGAGGAATCAGAGAAGACAGGCCAGCAACACACCTAGTCATATTCCTATCAGCAGTAGTTTCAGCACCAGTGTCTACCAACCAATTCCACAACCCACCACACCGGTTTCCTCCTTCACATCTGGCTCCATGTTGGGCCGAACAGACACAGCCCTCACAAACACCTACAGCGCTCTGCCGCCTATGCCCAGCTTCACCATGGCAAATA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Related Categories

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hassan Akrami et al.
Journal of ophthalmic & vision research, 4(3), 134-141 (2009-07-01)
To establish human retinal pigment epithelial (RPE) cell culture as a source for cell replacement therapy in ocular diseases. Human cadaver globes were used to isolate RPE cells. Each globe was cut into several pieces of a few millimeters in
Zhe Qian et al.
Respiratory research, 19(1), 262-262 (2018-12-31)
This study investigated the function of SMAD3 (SMAD family member 3) in regulating PAX6 (paired box 6) in non-small cell lung cancer. First, qRT-PCR was employed to detect SMAD3 expression in cancer tissues along with normal tissues and four cell lines
Yi Lu et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 129, 110381-110381 (2020-09-06)
Colorectal cancer is a kind of gastrointestinal tumor with rising morbidity and mortality. 5-fluorouracil is one of the most effective chemotherapy drugs for the treatment of CRC. However, clinical data reported dramatic resistance on the treatment for CRC with 5-fluorouracil.
Akira Ooki et al.
Oncogene, 37(45), 5967-5981 (2018-07-08)
It remains unclear whether PAX6 acts as a crucial transcription factor for lung cancer stem cell (CSC) traits. We demonstrate that PAX6 acts as an oncogene responsible for induction of cancer stemness properties in lung adenocarcinoma (LUAD). Mechanistically, PAX6 promotes
Zhengjia Liu et al.
American journal of translational research, 12(6), 2538-2553 (2020-07-14)
This article explored LINC01619 impact on non-small cell lung cancer (NSCLS) development. LINC01619 expression in tumor tissues/normal tissues of NSCLS patients was detected by qRT-PCR and in situ hybridization. PAX6 expression in clinical tissues was researched by immunohistochemistry. After transfection

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service