Skip to Content
Merck
All Photos(1)

Key Documents

EHU089961

Sigma-Aldrich

MISSION® esiRNA

targeting human LYN

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCAGAGGGAATGGCATACATCGAGCGGAAGAACTACATTCACCGGGACCTGCGAGCAGCTAATGTTCTGGTCTCCGAGTCACTCATGTGCAAAATTGCAGATTTTGGCCTTGCTAGAGTAATTGAAGATAATGAGTACACAGCAAGGGAAGGTGCTAAGTTCCCTATTAAGTGGACGGCTCCAGAAGCAATCAACTTTGGATGTTTCACTATTAAGTCTGATGTGTGGTCCTTTGGAATCCTCCTATACGAAATTGTCACCTATGGGAAAATTCCCTACCCAGGGAGAACTAATGCCGACGTGATGACCGCCCTGTCCCAGGGCTACAGGATGCCCCGTGTGGAGAACTGCCCAGATGAGCTCTATGACATTATGAAAATGTGCTGGAAAGAAAAGGCAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuaibin Liu et al.
Oncotarget, 7(46), 75468-75481 (2016-10-01)
Cervical cancer is one of the most common malignant tumor in women. The mechanisms of cervical cancer are intricate and have not been fully understood. Therefore, we employed iTRAQ to obtain novel proteins profile which participates in the tumor oncogenesis
Xiaoyun Wang et al.
Scientific reports, 7, 42675-42675 (2017-02-17)
Hypersecretion of mucus is an important component of airway remodeling and contributes to the mucus plugs and airflow obstruction associated with severe asthma phenotypes. Lyn has been shown to down-regulate allergen-induced airway inflammation. However, the role of Lyn in mucin
Lei Meng et al.
Acta biochimica et biophysica Sinica, 52(1), 49-57 (2019-12-13)
Gastric cancer (GC) is one of malignant tumors with high mortality and morbidity in the world. MicroRNA-122 (miR-122) acts as a tumor suppressor in a variety of cancers and has been found to be dominant in gastric adenocarcinoma. However, the
D Thaper et al.
Oncogene, 36(28), 3964-3975 (2017-03-14)
The acquisition of an invasive phenotype by epithelial cells occurs through a loss of cellular adhesion and polarity, heralding a multistep process that leads to metastatic dissemination. Since its characterization in 1995, epithelial-mesenchymal transition (EMT) has been closely linked to
Pradip Das et al.
Journal of immunology (Baltimore, Md. : 1950), 206(1), 181-192 (2020-12-06)
MCP-1-induced monocyte chemotaxis is a crucial event in inflammation and atherogenesis. Identifying the important signal transduction pathways that control monocyte chemotaxis can unravel potential targets for preventive therapies in inflammatory disease conditions. Previous studies have shown that the focal adhesion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service