Skip to Content
Merck
All Photos(1)

Documents

EHU088741

Sigma-Aldrich

MISSION® esiRNA

targeting human HDAC8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCCAGTATGGTGCATTCTTTGATTGAAGCATATGCACTGCATAAGCAGATGAGGATAGTTAAGCCTAAAGTGGCCTCCATGGAGGAGATGGCCACCTTCCACACTGATGCTTATCTGCAGCATCTCCAGAAGGTCAGCCAAGAGGGCGATGATGATCATCCGGACTCCATAGAATATGGGCTAGGTTATGACTGCCCAGCCACTGAAGGGATATTTGACTATGCAGCAGCTATAGGAGGGGCTACGATCACAGCTGCCCAATGCCTGATTGACGGAATGTGCAAAGTAGCAATTAACTGGTCTGGAGGGTGGCATCATGCAAAGAAAGATGAAGCATCTGGTTTTTGTTATCTCAATGATGCTGTCCTGGGAATATTACGATTGCGACGGAAATTTGAGCGTATTCTCTACGTGGATTTGGATCTGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael F Emmons et al.
Cancer research, 79(11), 2947-2961 (2019-04-17)
Melanoma cells have the ability to switch to a dedifferentiated, invasive phenotype in response to multiple stimuli. Here, we show that exposure of melanomas to multiple stresses including BRAF-MEK inhibitor therapy, hypoxia, and UV irradiation leads to an increase in
Zhouxiang Jin et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 84, 1645-1653 (2016-11-17)
Despite the use of novel anti-myeloma agents,nearly all patients will eventually relapse or become refractory to drug treatment. New and more effective drugs for multiple myeloma (MM) are urgently needed. The JAK-STAT signaling pathway is important in the proliferation of
Yunhe Zhang et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(6), 7295-7310 (2020-04-14)
Histone deacetylases (HDACs) have been shown to alleviate renal fibrosis, however, the role of individual HDAC isoforms in this process is poorly understood. In this study, we examined the role of HDAC8 in the development of renal fibrosis and partial
G R Vanaja et al.
Cell communication and signaling : CCS, 16(1), 20-20 (2018-05-03)
Histone deacetylases (HDACs) are involved in epigenetic gene regulation via deacetylation of acetylated lysine residues of both histone and non-histone proteins. Among the 18 HDACs identified in humans, HDAC8, a class I HDAC, is best understood structurally and enzymatically. However
Ganggang Kong et al.
Journal of neurotrauma, 34(18), 2645-2655 (2017-07-08)
The ketone metabolite β-hydroxybutyrate (βOHB), is reported to be neuroprotective after spinal cord injury (SCI) in rats, but the underlying mechanism remains unknown. The present study aims to investigate effects of βOHB on suppression of oxidative stress and inhibition of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service