Transfection protocols vary from transfection reagent and cell line. Transfect the cell line of interest with esiRNA following the manufacturer’s instructions for the transfection reagent. esiRNA should be tested in a pilot experiment to validate the best concentration and experimental procedure to use with every cell line. Known transfection conditions used for chemically synthesized siRNA are a good starting point for optimization.
Select a Size
$335.00
Select a Size
About This Item
$335.00
Recommended Products
description
Powered by Eupheria Biotech
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
AAGACCCGTGTGGTCAGCTACCGCGTGCCCCACAACGCTGCGGTGCAGGTGTACGACTACCGAGAGAAGCGAGCCCGCGTGGTCTTCGGGCCTGAGCTGGTGTCGCTGGGTCCTGAGGAGCAGTTCACAGTGTTGTCCCTCTCAGCTGGGCGGCCCAAGCGTCCCCATGCCCGCCGTGCGCTCTGCCTGCTGCTGGGGCCTGACTTCTTCACAGACGTCATCACCATCGAAACGGCGGATCATGCCAGGCTGCAACTGCAGCTGGCCTACAACTGGCACTTTGAGGTGAATGACCGGAAGGACCCCCAAGAGACGGCCAAGCTCTTTTCAGTGCCAGACTTTGTAGGTGATGCCTGCAAAGCCATCGCATCCCGGGTGCGGGGGGCCGTGGCCTCTGTCACTTTCGATGACTTCCATAAGAACTCAGCCCG
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Related Categories
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How long should I transfect cells with this product?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service