Skip to Content
Merck
All Photos(1)

Key Documents

EHU086811

Sigma-Aldrich

MISSION® esiRNA

targeting human SYT1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Estimated to ship on27 April 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Estimated to ship on27 April 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCCATATAGTGCTCTTTAGCCAGTATCTGTAAATACCTCAGTAATATGGGTCCTTTCATTTTTCCAGCCATGCATTCCTAACACAATTCAGTGGTACTTGGAATCCTGTTTTAATTTGCACAAATTTAAATGTAGAGAGCCCCTAAGTCCTTCATCATACCACTGCCCTCCAAATCTACTCTTCTTTTAAGCAATATGATGTGTAGATAGAGCATGAATGAAATTATTTATTGTATCACACTGTTGTATATACCAGTATGCTAAAGATTTATTTCTAGTTTGTGTATTTGTATGTTGTAAGCGTTTCCTAATCTGTGTATATCTAGATGTTTTTAATAAGATGTTCTATTTTAAACTATGTAAATTGACTGAGATATAGGAGAGCTGATAATATATTATACGGTAAATATAGTATCGTCTGCATTCCAGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng Xu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 46(2), 699-712 (2018-04-06)
Necroptosis, a form of programmed necrosis, is involved in the pathologic process of several kinds of pulmonary diseases. However, the role of necroptosis in particulate matter (PM)-induced pulmonary injury remains unclear. The objective of this study is to investigate the
Yang Xiao et al.
Biochimica et biophysica acta. Molecular basis of disease, 1864(5 Pt A), 1728-1743 (2018-02-25)
Diabetic cardiomyopathy is associated with suppressed autophagy and augmented inflammation in the heart. The effects of Tax1 binding protein 1 (TAX1BP1) on both autophagy and inflammation suggest that it may participate in the progression of diabetic cardiomyopathy. Mice were injected
Changping Gu et al.
Shock (Augusta, Ga.), 44(1), 83-89 (2015-03-24)
Recombinant human annexin A5 (Anx5) is known to protect cardiac function during endotoxemia, although the underlying mechanisms have yet to be elucidated. In this study, we demonstrated that Anx5 could repair the disrupted cardiomyocyte adherens junctions and improve the myocardial
Hirokuni Akahori et al.
Nature communications, 6, 7792-7792 (2015-08-06)
Macrophages are an essential component of the immune response to ischaemic injury and play an important role in promoting inflammation and its resolution, which is necessary for tissue repair. The type I transmembrane glycoprotein CD163 is exclusively expressed on macrophages
Y Loriot et al.
Cell death & disease, 5, e1423-e1423 (2014-09-19)
Radiotherapy has a critical role in the treatment of small-cell lung cancer (SCLC). The effectiveness of radiation in SCLC remains limited as resistance results from defects in apoptosis. In the current study, we investigated whether using the Bcl-2/Bcl-XL inhibitor S44563

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service