Skip to Content
Merck
All Photos(1)

Key Documents

EHU084891

Sigma-Aldrich

MISSION® esiRNA

targeting human SLC8A1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGTGGCCCTTACCATTATCCGCAGAGGTGGTGATTTGACTAACACTGTGTTTGTTGACTTCAGAACAGAGGATGGCACAGCAAATGCTGGGTCTGATTATGAATTTACTGAAGGAACTGTGGTGTTTAAGCCTGGTGATACCCAGAAGGAAATCAGAGTGGGTATCATAGATGATGATATCTTTGAGGAGGATGAAAATTTCCTTGTGCATCTCAGCAATGTCAAAGTATCTTCTGAAGCTTCAGAAGATGGCATACTGGAAGCCAATCATGTTTCTACACTTGCTTGCCTCGGATCTCCCTCCACTGCCACTGTAACTATTTTTGATGATGACCACGCAGGCATTTTTACTTTTGAGGAACCTGTGACTCATGTGAGTGAGAGCATTGGCATCATGGAGGTGAAAGTATTGAGAACATCTGGAGCTCGAGGAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guendalina Bastioli et al.
Cell death & disease, 10(2), 80-80 (2019-01-30)
Progressive accumulation of α-synuclein (α-syn) and exposure to environmental toxins are risk factors that may both concur to Parkinson's disease (PD) pathogenesis. Electrophysiological recordings of field postsynaptic potentials (fEPSPs) and Ca2+ measures in striatal brain slices and differentiated SH-SY5Y cells
Barbora Chovancova et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 44(2), 763-777 (2017-11-24)
Melatonin is a hormone transferring information about duration of darkness to the organism and is known to modulate several signaling pathways in the cells, e.g. generation of endoplasmic reticulum stress, oxidative status of the cells, etc. Melatonin has been shown
Silvia Piccirillo et al.
Cell death & disease, 9(7), 731-731 (2018-06-30)
In brain ischemia, reduction in oxygen and substrates affects mitochondrial respiratory chain and aerobic metabolism, culminating in ATP production impairment, ionic imbalance, and cell death. The restoration of blood flow and reoxygenation are frequently associated with exacerbation of tissue injury
M Song et al.
British journal of pharmacology, 171(14), 3432-3447 (2014-03-20)
SKF 96365 is well known for its suppressing effect on human glioblastoma growth by inhibiting pre-activated transient receptor potential canonical (TRPC) channels and Ca(2+) influx. The effect of SKF 96363 on glioblastoma cells, however, may be multifaceted and this possibility

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service