Skip to Content
Merck
All Photos(1)

Documents

EHU084101

Sigma-Aldrich

MISSION® esiRNA

targeting human TGFB2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGCACCTCCACATATACCAGTGGTGATCAGAAAACTATAAAGTCCACTAGGAAAAAAAACAGTGGGAAGACCCCACATCTCCTGCTAATGTTATTGCCCTCCTACAGACTTGAGTCACAACAGACCAACCGGCGGAAGAAGCGTGCTTTGGATGCGGCCTATTGCTTTAGAAATGTGCAGGATAATTGCTGCCTACGTCCACTTTACATTGATTTCAAGAGGGATCTAGGGTGGAAATGGATACACGAACCCAAAGGGTACAATGCCAACTTCTGTGCTGGAGCATGCCCGTATTTATGGAGTTCAGACACTCAGCACAGCAGGGTCCTGAGCTTATATAATACCATAAATCCAGAAGCATCTGCTTCTCCTTGCTGCGTGTCCCAAGATTTAGAACCTCTAACCATTCTCTACTACATTGGCAAAACACCCAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph Yusup Shin et al.
Science translational medicine, 11(497) (2019-06-21)
In systemic sclerosis (SSc), previously healthy adults develop an inflammatory prodrome with subsequent progressive fibrosis of the skin and viscera. SSc has a weak signature for genetic contribution, and there are few pathogenic insights or targeted treatments for this condition.
Ruijing Lu et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(9), 6691-6700 (2015-03-31)
A large body of evidence indicates that microRNAs play a critical role in tumor initiation and progression by negatively regulating oncogenes or tumor suppressor genes. Here, we report that the expression of miR-200a was notably downregulated in 45 renal cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service