Skip to Content
Merck
All Photos(1)

Documents

EHU075951

Sigma-Aldrich

MISSION® esiRNA

targeting human FBXW7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGCAGTCACAGGCAAATGTCTGAGAACATTAGTGGGACATACAGGTGGAGTATGGTCATCACAAATGAGAGACAACATCATCATTAGTGGATCTACAGATCGGACACTCAAAGTGTGGAATGCAGAGACTGGAGAATGTATACACACCTTATATGGGCATACTTCCACTGTGCGTTGTATGCATCTTCATGAAAAAAGAGTTGTTAGCGGTTCTCGAGATGCCACTCTTAGGGTTTGGGATATTGAGACAGGCCAGTGTTTACATGTTTTGATGGGTCATGTTGCAGCAGTCCGCTGTGTTCAATATGATGGCAGGAGGGTTGTTAGTGGAGCATATGATTTTATGGTAAAGGTGTGGGATCCAGAGACTGAAACCTGTCTACACACGTTGCAGGGGCATACTAAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariusz L Hartman et al.
Molecular carcinogenesis, 58(4), 588-602 (2018-12-18)
We have extensively studied the phenotypic heterogeneity of patient-derived melanoma cells. Here, whole-exome sequencing revealed novel variants of genes associated with the MAPK, NOTCH, Hippo, cell-cycle, senescence, and ubiquitin-dependent pathways, which could contribute to the observed phenotypic diversity between cell
Qiang Fu et al.
Aging, 11(24), 11865-11879 (2019-12-28)
Our study aspires to understand the impact of miR-27b on myocardial fibrosis as well as its functional mechanism. 12 days post the ligation of coronary artery in rats, the expression of miR-27b in the peri-infarction region was elevated. Treating cultivated
Veronika Reiterer et al.
The EMBO journal, 36(3), 260-273 (2016-12-23)
The F-box protein FBXW7 is the substrate-recruiting subunit of an SCF ubiquitin ligase and a major tumor-suppressor protein that is altered in several human malignancies. Loss of function of FBXW7 results in the stabilization of numerous proteins that orchestrate cell
Wei Huo et al.
Cancer cell international, 20, 64-64 (2020-03-07)
Dysregulation of long non-coding RNAs (lncRNAs) is involved in development of prostate cancer. However, the molecular mechanisms of many lncRNAs in prostate cancer have not been studied yet. The lncRNA Fer-1-like protein 4 (FER1L4) expression was explored in prostate tumors
Shusaku Honma et al.
Cancers, 11(5) (2019-05-10)
Although the cancer stem cell (CSC) concept has provided a reasonable explanation for cancer recurrence following chemotherapy, the relationship between CSCs and chemotherapy resistance has not been thoroughly investigated, especially in solid tumors. We aimed to identify the mechanism underlying

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service