Skip to Content
Merck
All Photos(1)

Key Documents

EHU075391

Sigma-Aldrich

MISSION® esiRNA

targeting human GREM1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Check Cart for Availability


Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Check Cart for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AAGCGAGACTGGTGCAAAACCCAGCCGCTTAAGCAGACCATCCACGAGGAAGGCTGCAACAGTCGCACCATCATCAACCGCTTCTGTTACGGCCAGTGCAACTCTTTCTACATCCCCAGGCACATCCGGAAGGAGGAAGGTTCCTTTCAGTCCTGCTCCTTCTGCAAGCCCAAGAAATTCACTACCATGATGGTCACACTCAACTGCCCTGAACTACAGCCACCTACCAAGAAGAAGAGAGTCACACGTGTGAAGCAGTGTCGTTGCATATCCATCGATTTGGATTAAGCCAAATCCAGGTGCACCCAGCATGTCCTAGGAATGCAGCCCCAGGAAGTCCCAGACCTAAAACAACCAGATTCTTACTTGGCTTAAACCTAGAGGCCAGAAGAACCCCCAGCTGCCTCCTGGCAGGAGCCTGCTTGTGCGTAGTTCGTGTGCATGAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duo Li et al.
Molecular vision, 25, 625-635 (2019-11-09)
To investigate the role of Gremlin-1, which is an endogenous antagonist of the bone morphogenetic protein (BMP) signaling pathway, in inducing epithelium-mesenchymal transition (EMT) in fetal RPE cells after repeated wounds. Subconfluent repetitive passages in fetal RPE cells were regarded
Nam Ji Sung et al.
International journal of molecular sciences, 21(23) (2020-12-09)
Gremlin-1 (GREM1), one of the bone morphogenetic protein (BMP) antagonists, can directly bind to BMPs. GREM1 is involved in organogenesis, tissue differentiation, and organ fibrosis. Recently, numerous studies have reported the oncogenic role of GREM1 in cancer. However, the role
Na Hui Kim et al.
Biochemical and biophysical research communications, 533(4), 1378-1384 (2020-10-25)
Gremlin-1 (GREM1), one of the antagonists of bone morphogenetic proteins (BMPs), has recently been reported to be overexpressed in a variety of cancers including breast cancer. GREM1 is involved in tumor promotion, but little is known about its role in
Kongzu Hu et al.
Molecular medicine reports, 15(4), 2186-2194 (2017-03-06)
Previous research focusing on rodent cells and animal models has demonstrated that gremlin-1 antagonizes bone morphogenetic proteins (BMPs) in order to suppress osteogenesis. However, the impact of gremlin‑1 on osteogenesis in human bone marrow-derived mesenchymal stem cells (MSCs) remains unknown.
Julie R Graham et al.
Journal of cellular biochemistry, 115(9), 1539-1548 (2014-03-19)
Fibrosis is a chronic disease characterized by an excessive deposition of scar tissue in the affected organs. A central mediator of this process is transforming growth factor-β (TGF-β), which stimulates the production of extracellular matrix proteins such as collagens. MicroRNAs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service