Skip to Content
Merck
All Photos(1)

Key Documents

EHU073351

Sigma-Aldrich

MISSION® esiRNA

targeting human ACTB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTCCACCTTCCAGCAGATGTGGATCAGCAAGCAGGAGTATGACGAGTCCGGCCCCTCCATCGTCCACCGCAAATGCTTCTAGGCGGACTATGACTTAGTTGCGTTACACCCTTTCTTGACAAAACCTAACTTGCGCAGAAAACAAGATGAGATTGGCATGGCTTTATTTGTTTTTTTTGTTTTGTTTTGGTTTTTTTTTTTTTTTTGGCTTGACTCAGGATTTAAAAACTGGAACGGTGAAGGTGACAGCAGTCGGTTGGAGCGAGCATCCCCCAAAGTTCACAATGTGGCCGAGGACTTTGATTGCACATTGTTGTTTTTTTAATAGTCATTCCAAATATGAGATGCGTTGTTACAGGAAGTCCCTTGCCATCCTAAAAGCCACCCCACTTCTCTCTAAGGAGAATGGCCCAGTCCTCTCCCAAGTCCACACAGGGGAGGTGATAGCAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... ACTB(60) , ACTB(60)

Related Categories

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katariina Nurmi et al.
PloS one, 8(11), e78537-e78537 (2013-11-19)
In the pathogenesis of coronary atherosclerosis, local macrophage-driven inflammation and secretion of proinflammatory cytokines, interleukin-1β (IL-1β) in particular, are recognized as key factors. Moderate alcohol consumption is associated with a reduced risk of coronary artery disease mortality. Here we examined
Dimitris Stellas et al.
BMC cell biology, 11, 51-51 (2010-07-07)
Heat shock protein 90 (HSP90) is a molecular chaperone that is considered a new target for the treatment of cancer. Increasing data reveal an extracellular chaperoning activity for HSP90. Here we investigate the interaction of the secreted isoforms of HSP90
Isabella W Y Mak et al.
Journal of cellular biochemistry, 116(1), 37-44 (2014-08-26)
Chondrosarcoma is the most common primary bone malignancy in the adult population with a high rate of pulmonary metastasis. Chondrosarcoma is managed with surgical excision as the tumors do not respond well to conventional chemotherapy or radiation therapy. Thus, there
Lesley A Everett et al.
Blood, 123(24), 3697-3705 (2014-04-11)
The primary cellular source of factor VIII (FVIII) biosynthesis is controversial, with contradictory evidence supporting an endothelial or hepatocyte origin. LMAN1 is a cargo receptor in the early secretory pathway that is responsible for the efficient secretion of factor V
Antonella Spagnoletti et al.
Molecular and cellular endocrinology, 412, 56-64 (2015-06-02)
Bisphenol A (BPA) and para-Nonylphenol (p-NP) are chemicals of industrial origin which may influence human reproductive health. The effects of these substances in the prenatal life is an important topic that is receiving greater attention in the developed countries. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service