Skip to Content
Merck
All Photos(1)

Key Documents

EHU070211

Sigma-Aldrich

MISSION® esiRNA

targeting human TRPC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TTTGGAAAATTTCTTGGGATGTTTCTTCTTGTTTTGTTTTCTTTCACAATTGGACTGACACAACTGTATGATAAAGGATATACTTCAAAGGAGCAGAAGGACTGTGTAGGCATCTTCTGTGAACAGCAAAGCAATGATACCTTCCATTCGTTCATTGGCACCTGCTTTGCTTTGTTCTGGTATATTTTCTCCTTAGCGCATGTGGCAATCTTTGTCACAAGATTTAGCTATGGAGAAGAACTGCAGTCCTTTGTGGGAGCTGTCATTGTTGGTACATACAATGTCGTGGTTGTGATTGTGCTTACCAAACTGCTGGTGGCAATGCTTCATAAAAGCTTTCAGTTGATAGCAAATCATGAAGACAAAGAATGGAAGTTTGCTCGAGCAAAATTATGGCTTAGCTACTTTGATGACAAATGTACGTTACCTCCACCTTTCAACATCATTCCCTCACCA

Ensembl | human accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinqin Pu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(1), 1074-1085 (2018-08-02)
Airway remodeling with progressive epithelial alterations in the respiratory tract is a severe consequence of asthma. Although dysfunctional signaling transduction is attributed to airway inflammation, the exact mechanism of airway remodeling remains largely unknown. TRPC1, a member of the transient
DongXu He et al.
Science advances, 6(12), eaaz3367-eaaz3367 (2020-03-25)
Mammalian transient receptor potential (TRP) channels are major components of Ca2+ signaling pathways and control a diversity of physiological functions. Here, we report a specific role for TRPC1 in the entry of herpes simplex virus type 1 (HSV-1) into cells.
Corena V Grant et al.
Breast cancer research and treatment, 177(2), 345-355 (2019-06-24)
Triple-negative breast cancers (TNBCs) represent a heterogeneous group of tumors. The lack of targeted therapies combined with the inherently aggressive nature of TNBCs results in a higher relapse rate and poorer overall survival. We evaluated the heterogeneity of TNBC cell
Xiaoyu Zhang et al.
Journal of cellular biochemistry, 119(7), 6033-6044 (2018-03-27)
This study aimed to validate whether transient receptor potential channel1 (TRPC1) and TRPC3 participate in the regulation the proliferation of airway smooth muscle cells (ASMCs) through modulating calcium ion (Ca2+ ) influx in vitro. Chronic model of murine asthma was
Yu Fu et al.
Cells, 10(1) (2021-01-17)
In animals, muscle growth is a quantitative trait controlled by multiple genes. Previously, we showed that the transient receptor potential channel 1 (TRPC1) gene was differentially expressed in muscle tissues between pig breeds with divergent growth traits base on RNA-seq.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service