Skip to Content
Merck
All Photos(1)

Key Documents

EHU063971

Sigma-Aldrich

MISSION® esiRNA

targeting human CALCR

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Estimated to ship on18 April 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Estimated to ship on18 April 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCCTGCAACTATTTCTGGATGCTCTGTGAAGGGATCTATCTTCATACACTCATTGTCGTGGCTGTGTTTACTGAGAAGCAACGCTTGCGGTGGTATTATCTCTTGGGCTGGGGGTTCCCGCTGGTGCCAACCACTATCCATGCTATTACCAGGGCCGTGTACTTCAATGACAACTGCTGGCTGAGTGTGGAAACCCATTTGCTTTACATAATCCATGGACCTGTCATGGCGGCACTTGTGGTCAATTTCTTCTTTTTGCTCAACATTGTCCGGGTGCTTGTGACCAAAATGAGGGAAACCCATGAGGCGGAATCCCACATGTACCTGAAGGCTGTGAAGGCCACCATGATCCTTGTGCCCCTGCTGGGAATCCAGTTTGTCGTCTTTCCCTGGAGACCTTCCAACAAGATGCTTGGGAAGATATATGATTACGTGATGCACTCTCTGATTCATTTCCAGGGCTTCTTTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Toffoli et al.
Clinical science (London, England : 1979), 134(17), 2337-2352 (2020-08-29)
TNF-related apoptosis-inducing ligand (TRAIL) has attracted attention not only as an anti-cancer agent, but also as a potential treatment for diabetes. Animal studies have shown that TRAIL delivery ameliorated glucose control in type 1 and type 2 diabetes. It is
Ruo Feng et al.
Diagnostic pathology, 10, 149-149 (2015-08-27)
Hepatocellular carcinoma (HCC) is one of the most frequent cancers in the world. Calreticulin(CRT) is aberrantly overexpressed in many human cancer cells. The function of CRT in HCC cells remains unclear. We attempted to investigate the effects and the underlying
Xuemin Wang et al.
PloS one, 10(4), e0125402-e0125402 (2015-05-01)
Cisplatin is one of the first-line platinum-based chemotherapeutic agents for treatment of many types of cancer, including ovary cancer. CTR1 (copper transporter 1), a transmembrane solute carrier transporter, has previously been shown to increase the cellular uptake and sensitivity of
Saurabh Vig et al.
Cell cycle (Georgetown, Tex.), 14(14), 2274-2284 (2015-05-07)
Calreticulin (CRT) is an endoplasmic reticulum (ER) resident calcium binding protein that is involved in several cellular activities. Transcriptome analyses in CRT knockdown HepG2 cells revealed 253 altered unique genes and subsequent in silico protein-protein interaction network and MCODE clustering
Beatrice Rondinelli et al.
Nucleic acids research, 43(5), 2560-2574 (2015-02-26)
DNA replication is a tightly regulated process that initiates from multiple replication origins and leads to the faithful transmission of the genetic material. For proper DNA replication, the chromatin surrounding origins needs to be remodeled. However, remarkably little is known

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service