Skip to Content
Merck
All Photos(1)

Key Documents

EHU060511

Sigma-Aldrich

MISSION® esiRNA

targeting human EPHB2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCACACCCAGTACACCTTCGAGATCCAGGCTGTGAACGGCGTTACTGACCAGAGCCCCTTCTCGCCTCAGTTCGCCTCTGTGAACATCACCACCAACCAGGCAGCTCCATCGGCAGTGTCCATCATGCATCAGGTGAGCCGCACCGTGGACAGCATTACCCTGTCGTGGTCCCAGCCGGACCAGCCCAATGGCGTGATCCTGGACTATGAGCTGCAGTACTATGAGAAGGAGCTCAGTGAGTACAACGCCACAGCCATAAAAAGCCCCACCAACACGGTCACCGTGCAGGGCCTCAAAGCCGGCGCCATCTATGTCTTCCAGGTGCGGGCACGCACCGTGGCAGGTTACGGGCGCTACAGCGGCAAGATGTACTTCCAGACCATGACAGAAGCCGAGTACCAGACAAGCATCCAGGAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiuqing Li et al.
PloS one, 9(8), e105326-e105326 (2014-08-26)
Effective treatment of transitional cell carcinoma (TCC) of the bladder requires early diagnosis. Identifying novel molecular markers in TCC would guide the development of diagnostic and therapeutic targets. Ephrins mediate signals via tyrosine kinase activity that modulates diverse physiologic and
Walaiporn Khansaard et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(10), 10031-10041 (2014-07-12)
The activation of Ephrin (Eph) receptors, the largest tyrosine kinase families of cell surface receptor, has recently been addressed in human cholangiocarcinoma (CCA). Therefore, the present study aimed to investigate the role of Eph receptors and its ligands in CCA.
Young Hyun Jung et al.
Biochimica et biophysica acta, 1853(8), 1905-1917 (2015-05-13)
The role of unsaturated fatty acids (UFAs) is essential for determining stem cell functions. Eph/Ephrin interactions are important for regulation of stem cell fate and localization within their niche, which is significant for a wide range of stem cell behavior.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service