Skip to Content
Merck
All Photos(1)

Key Documents

EHU058001

Sigma-Aldrich

MISSION® esiRNA

targeting human CLU

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Estimated to ship on09 April 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Estimated to ship on09 April 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACTCTGCTGCTGTTTGTGGGGCTGCTGCTGACCTGGGAGAGTGGGCAGGTCCTGGGGGACCAGACGGTCTCAGACAATGAGCTCCAGGAAATGTCCAATCAGGGAAGTAAGTACGTCAATAAGGAAATTCAAAATGCTGTCAACGGGGTGAAACAGATAAAGACTCTCATAGAAAAAACAAACGAAGAGCGCAAGACACTGCTCAGCAACCTAGAAGAAGCCAAGAAGAAGAAAGAGGATGCCCTAAATGAGACCAGGGAATCAGAGACAAAGCTGAAGGAGCTCCCAGGAGTGTGCAATGAGACCATGATGGCCCTCTGGGAAGAGTGTAAGCCCTGCCTGAAACAGACCTGCATGAAGTTCTACGCACGCGTCTGCAGAAGTGGCTCAGGCCTGGTTGGCCGCCAGCTTGAGGAGTTCCTGAACCAGAGCTCGCCCTTCTAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na Fu et al.
Life sciences, 256, 117911-117911 (2020-06-07)
To explore the potential regulatory mechanism of differentially expressed mRNAs in Hepatitis C virus (HCV)-related hepatocellular carcinoma (HCC). Patients with HCV-related HCC and age- and gender-matched healthy subjects were enrolled. Differentially expressed mRNAs in the plasma were detected by digital
Lihua Mu et al.
Bioengineered, 11(1), 472-483 (2020-04-07)
Recent focus has turned to secretory clusterin (sCLU) as a key contributor to chemoresistance of anticancer agents, but the role of sCLU on chemotherapy drug response to gastric cancer cells is not fully understood. Previous research found that sCLU was
Chiara Tarquini et al.
Aging, 12(11), 10129-10146 (2020-06-10)
Osteoarthritis (OA) is the most common joint disease characterized by destruction of articular cartilage. OA-induced cartilage degeneration causes inflammation, oxidative stress and the hypertrophic shift of quiescent chondrocytes. Clusterin (CLU) is a ubiquitous glycoprotein implicated in many cellular processes and
Xiaohui Wang et al.
Current pharmaceutical biotechnology, 21(2), 131-139 (2019-08-23)
The aim of the study was to investigate the expression of sCLU in relation to the clinicopathological features and prognosis of patients with untreated High-Grade Osteosarcoma (HGOS) and to evaluate sCLU as a target for osteosarcoma (OS) therapies. The expression
Jung-Yoon Heo et al.
The Journal of endocrinology, 237(2), 175-191 (2018-03-23)
Clusterin is a secretory glycoprotein that is involved in multiple physiopathological processes, including lipid metabolism. Previous studies have shown that clusterin prevents hepatic lipid accumulation via suppression of sterol regulatory element-binding protein (SREBP) 1. In this study, we examined the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service