Skip to Content
Merck
All Photos(1)

Key Documents

EHU055111

Sigma-Aldrich

MISSION® esiRNA

targeting human NOTCH3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$335.00
50 μG
$584.00

$335.00


Estimated to ship on16 May 2025



Select a Size

Change View
20 μG
$335.00
50 μG
$584.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

$335.00


Estimated to ship on16 May 2025


description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCTCAGACTGGTCCGAATCCACGCCTAGCCCAGCCACTGCCACTGGGGCCATGGCCACCACCACTGGGGCACTGCCTGCCCAGCCACTTCCCTTGTCTGTTCCCAGCTCCCTTGCTCAGGCCCAGACCCAGCTGGGGCCCCAGCCGGAAGTTACCCCCAAGAGGCAAGTGTTGGCCTGAGACGCTCGTCAGTTCTTAGATCTTGGGGGCCTAAAGAGACCCCCGTCCTGCCTCCTTTCTTTCTCTGTCTCTTCCTTCCTTTTAGTCTTTTTCATCCTCTTCTCTTTCCACCAACCCTCCTGCATCCTTGCCTTGCAGCGTGACCGAGATAGGTCATCAGCCCAGGGCTTCAGTCTTCCTTTATTTATAATGGGTGGGGGCTACCACCCACCCTCTCAGTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yumi Yamamoto et al.
Molecular brain, 13(1), 38-38 (2020-03-20)
Cerebral autosomal dominant arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL) is one of the most common forms of hereditary cerebral small vessel diseases and is caused by mutations in NOTCH3. Our group has previously reported incorporation of NOTCH3 extracellular domain
Yash R Somnay et al.
Cancer, 123(5), 769-782 (2016-11-20)
Thyroid tumorigenesis is characterized by a progressive loss of differentiation exhibited by a range of disease variants. The Notch receptor family (1-4) regulates developmental progression in both normal and cancerous tissues. This study sought to characterize the third Notch isoform
Keito Okazaki et al.
Nature communications, 11(1), 5911-5911 (2020-11-22)
Transcriptional dysregulation, which can be caused by genetic and epigenetic alterations, is a fundamental feature of many cancers. A key cytoprotective transcriptional activator, NRF2, is often aberrantly activated in non-small cell lung cancers (NSCLCs) and supports both aggressive tumorigenesis and
Wei Hu et al.
Cancer research, 74(12), 3282-3293 (2014-04-20)
The Notch pathway plays an important role in the growth of high-grade serous ovarian (HGS-OvCa) and other cancers, but its clinical and biologic mechanisms are not well understood. Here, we found that the Notch pathway alterations are prevalent and significantly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service