Skip to Content
Merck
All Photos(1)

Documents

EHU054421

Sigma-Aldrich

MISSION® esiRNA

targeting human NACC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TCCCGGCTGAACTTATCAACCAGATTGGGAACCGCTGCCACCCCAAGCTCTACGACGAGGGCGACCCCTCTGAGAAGCTGGAGCTGGTGACAGGCACCAACGTGTACATCACAAGGGCGCAGCTGATGAACTGCCACGTCAGCGCAGGCACGCGGCACAAGGTCCTACTGCGGCGGCTCCTGGCCTCCTTCTTTGACCGGAACACGCTGGCCAACAGCTGCGGCACCGGCATCCGCTCTTCTACCAACGATCCCCGTCGGAAGCCCCTGGACAGCCGCGTGCTCCACGCTGTCAAGTACTACTGCCAGAACTTCGCCCCCAACTTCAAGGAGAGCGAGATGAATGCCATCGCGGCCGACATGTGCACCAACGCCCGCCGCGTCGTGCGCAAGAGCTGGATGCCCAAGGTCAAGGTGCTCAAGGCTGAGGATGACGCCTACACCACCTTC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong-Fang Tao et al.
Oncology reports, 45(2), 469-480 (2021-01-09)
Long non‑coding RNA (lncRNA) forkhead box P4 antisense RNA 1 (FOXP4‑AS1) has been determined to function as an oncogene in various types of cancer. However, the biological function and the underlying mechanisms of FOXP4‑AS1 in mantle cell lymphoma (MCL) remain to be uncovered. The
Kohei Morita et al.
Cancers, 10(10) (2018-09-27)
The nucleus accumbens-associated protein 1 (NACC1) is a transcription factor constitutively expressed in the urothelium, where it regulates cell growth, senescence, autophagy, and epithelial-mesenchymal transition. microRNA (miRNA) constitutes a class of small non-coding RNAs which are involved in cell proliferation
Xiao-Han Tang et al.
Cancer medicine, 8(14), 6426-6436 (2019-09-07)
Heparin-binding epidermal growth factor-like growth factor (HB-EGF) is a new promising target for the treatment of ovarian cancer. Our previous study showed that cross-reacting material 197 (CRM197), a specific HB-EGF inhibitor, significantly reverses resistance against paclitaxel in paclitaxel-resistant ovarian cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service